Assay Design



Gene Transcripts

Gene Symbol
ALKBH6
Entrez Gene ID
84964
Gene Name
alkB homolog 6
Gene Aliases
ABH6
Chromosome Location
Chr.19: 36009116 - 36014243 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001297701.1 NP_001284630.1 7 - 8 626 78
NM_032878.3 NP_116267.3 6 - 7 547 78
NM_198867.1 NP_942567.1 6 - 7 509 78
XM_005259357.4 XP_005259414.1 6 - 7 594 78
GenBank mRNA AB273714.1 - 7 - 8 626 78
AK304805.1 - 6 - 7 483 78
AK310704.1 - 5 - 6 786 78
AK314150.1 - 6 - 7 526 78
AK315894.1 - 5 - 6 725 78
BC007601.1 - 6 - 7 548 78
BC105801.1 - 6 - 7 519 78

View Full Product Details