Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132659971-132660465 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL13
Entrez Gene ID
3596
Gene Name
interleukin 13
Gene Aliases
IL-13, P600
UniGene
Hs.845
Gene Chromosome Location
Chr.5: 132658173-132661109 on Build GRCh38
Transcripts
NM_002188.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5021482 c.334-7C>T p.? Chr.5: 132660168-132660168 on Build GRCh38
7218663 c.431A>G p.Q144R Chr.5: 132660272-132660272 on Build GRCh38
232530 c.383T>G p.F128C Chr.5: 132660224-132660224 on Build GRCh38
3373659 c.372G>A p.E124E Chr.5: 132660213-132660213 on Build GRCh38
1210486 c.430C>T p.Q144* Chr.5: 132660271-132660271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775363860 Chr.5: 132660299-132660299 on Build GRCh38
rs560547280 Chr.5: 132659997-132659997 on Build GRCh38
rs765294353 Chr.5: 132660160-132660160 on Build GRCh38
rs374830989 Chr.5: 132660084-132660087 on Build GRCh38
rs552103568 Chr.5: 132660098-132660098 on Build GRCh38
rs751214731 Chr.5: 132660129-132660129 on Build GRCh38
rs566039790 Chr.5: 132660074-132660074 on Build GRCh38
rs763882844 Chr.5: 132660271-132660271 on Build GRCh38
rs376384925 Chr.5: 132660208-132660208 on Build GRCh38
rs199797009 Chr.5: 132660264-132660264 on Build GRCh38
rs569385458 Chr.5: 132660318-132660318 on Build GRCh38
rs2069749 Chr.5: 132660319-132660319 on Build GRCh38
rs2069748 Chr.5: 132660121-132660121 on Build GRCh38
rs200380202 Chr.5: 132660234-132660234 on Build GRCh38
rs1053570408 Chr.5: 132660102-132660102 on Build GRCh38
rs756919832 Chr.5: 132660196-132660196 on Build GRCh38
rs199969967 Chr.5: 132660126-132660126 on Build GRCh38
rs375653129 Chr.5: 132660211-132660211 on Build GRCh38
rs201475069 Chr.5: 132660262-132660262 on Build GRCh38
rs531513937 Chr.5: 132660130-132660130 on Build GRCh38
rs20541 Chr.5: 132660272-132660272 on Build GRCh38
rs569085982 Chr.5: 132660043-132660043 on Build GRCh38
rs886759323 Chr.5: 132660120-132660120 on Build GRCh38
rs755393562 Chr.5: 132660289-132660289 on Build GRCh38
rs200797041 Chr.5: 132660378-132660378 on Build GRCh38
rs780405199 Chr.5: 132660131-132660131 on Build GRCh38
rs75868517 Chr.5: 132660333-132660333 on Build GRCh38
rs140196099 Chr.5: 132660214-132660214 on Build GRCh38
rs200786939 Chr.5: 132660442-132660442 on Build GRCh38
rs200196698 Chr.5: 132660437-132660437 on Build GRCh38
rs372270964 Chr.5: 132660136-132660136 on Build GRCh38
rs56318025 Chr.5: 132660325-132660325 on Build GRCh38
rs368320626 Chr.5: 132660128-132660128 on Build GRCh38
rs201802470 Chr.5: 132660436-132660436 on Build GRCh38
rs776578550 Chr.5: 132660152-132660152 on Build GRCh38
rs936095335 Chr.5: 132660083-132660083 on Build GRCh38
rs78628956 Chr.5: 132660210-132660210 on Build GRCh38
rs561356790 Chr.5: 132660150-132660150 on Build GRCh38
rs114654496 Chr.5: 132660140-132660140 on Build GRCh38
rs538082433 Chr.5: 132660076-132660076 on Build GRCh38
rs767198237 Chr.5: 132660288-132660288 on Build GRCh38
rs773364000 Chr.5: 132660148-132660148 on Build GRCh38
rs778362054 Chr.5: 132660015-132660015 on Build GRCh38
rs754553478 Chr.5: 132660059-132660059 on Build GRCh38
rs202047364 Chr.5: 132660141-132660141 on Build GRCh38
rs201624646 Chr.5: 132660231-132660231 on Build GRCh38
rs200247901 Chr.5: 132660267-132660267 on Build GRCh38
rs140828306 Chr.5: 132660197-132660197 on Build GRCh38
rs917475040 Chr.5: 132660052-132660052 on Build GRCh38
rs56258826 Chr.5: 132660278-132660278 on Build GRCh38
rs868100758 Chr.5: 132660037-132660037 on Build GRCh38
rs1005303103 Chr.5: 132660155-132660155 on Build GRCh38
rs201863406 Chr.5: 132660337-132660337 on Build GRCh38
rs758955797 Chr.5: 132660302-132660302 on Build GRCh38
rs1044563461 Chr.5: 132660075-132660075 on Build GRCh38
rs961082728 Chr.5: 132660166-132660166 on Build GRCh38
rs1050067483 Chr.5: 132660377-132660377 on Build GRCh38
rs201839307 Chr.5: 132660173-132660173 on Build GRCh38
rs1015724931 Chr.5: 132660158-132660158 on Build GRCh38
rs771297548 Chr.5: 132660257-132660257 on Build GRCh38
rs1295686 Chr.5: 132660151-132660151 on Build GRCh38
rs201181769 Chr.5: 132660170-132660170 on Build GRCh38
rs777911950 Chr.5: 132660215-132660215 on Build GRCh38
rs532383493 Chr.5: 132660064-132660064 on Build GRCh38
rs975205774 Chr.5: 132660017-132660017 on Build GRCh38
rs374535004 Chr.5: 132660294-132660294 on Build GRCh38
rs184944562 Chr.5: 132660191-132660191 on Build GRCh38
rs761274909 Chr.5: 132660277-132660277 on Build GRCh38
rs199516826 Chr.5: 132660369-132660369 on Build GRCh38
rs775360450 Chr.5: 132660265-132660265 on Build GRCh38
rs745694958 Chr.5: 132660263-132660263 on Build GRCh38
rs558907104 Chr.5: 132660298-132660298 on Build GRCh38
rs747026798 Chr.5: 132660228-132660228 on Build GRCh38

View Full Product Details