Assay Design



Gene Transcripts

Gene Symbol
GPD2
Entrez Gene ID
2820
Gene Name
glycerol-3-phosphate dehydrogenase 2
Gene Aliases
GDH2, GPDM, mGPDH
Chromosome Location
Chr.2: 156435290 - 156586403 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000408.4 NP_000399.3 1 - 2 130 103
GenBank mRNA AK292817.1 - 1 - 2 129 103
AL699359.1 - 1 - 2 130 103
U12424.1 - 1 - 2 119 103

View Full Product Details