Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000952.4 | NP_000943.1 | 2 - 2 | 898 | 98 |
NM_001164721.1 | NP_001158193.1 | 3 - 3 | 842 | 98 | |
NM_001164722.2 | NP_001158194.1 | 2 - 2 | 760 | 98 | |
NM_001164723.2 | NP_001158195.1 | 3 - 3 | 969 | 98 | |
GenBank mRNA | AK290240.1 | - | 2 - 2 | 644 | 98 |
AK292053.1 | - | 3 - 3 | 842 | 98 | |
AK313174.1 | - | 2 - 2 | 629 | 98 | |
AY275466.1 | - | 1 - 1 | 507 | 98 | |
BC013816.2 | - | 1 - 1 | 547 | 98 | |
BC063000.1 | - | 2 - 2 | 634 | 98 | |
BT009801.1 | - | 1 - 1 | 507 | 98 | |
CD013879.1 | - | 3 - 3 | 698 | 98 | |
D10202.1 | - | 2 - 2 | 619 | 98 | |
L07334.1 | - | 1 - 1 | 507 | 98 | |
M76674.1 | - | 1 - 1 | 532 | 98 | |
M80436.1 | - | 2 - 2 | 773 | 98 | |
S52624.1 | - | 2 - 2 | 698 | 98 |