Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001133.2 | NP_001124.1 | 7 - 8 | 872 | 116 |
XM_017007842.1 | XP_016863331.1 | 7 - 8 | 877 | 116 | |
XM_017007843.1 | XP_016863332.1 | 7 - 8 | 877 | 116 | |
XM_017007844.1 | XP_016863333.1 | 7 - 8 | 877 | 116 | |
GenBank mRNA | AK290556.1 | - | 7 - 8 | 872 | 116 |
BC109020.1 | - | 7 - 8 | 870 | 116 | |
BC109021.1 | - | 7 - 8 | 870 | 116 |