Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001912.4 | NP_001903.1 | 1 - 2 | 342 | 88 |
XM_011518263.1 | XP_011516565.1 | 1 - 2 | 882 | 88 | |
GenBank mRNA | AK055599.1 | - | 1 - 2 | 882 | 88 |
BC012612.1 | - | 1 - 2 | 190 | 88 | |
BI829385.1 | - | 1 - 2 | 296 | 88 | |
X12451.1 | - | 1 - 2 | 280 | 88 |