Assay Design



Gene Transcripts

Gene Symbol
MKI67
Entrez Gene ID
4288
Gene Name
marker of proliferation Ki-67
Gene Aliases
KIA, MIB-, MIB-1, PPP1R105
Chromosome Location
Chr.10: 128096659 - 128126405 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145966.1 NP_001139438.1 6 - 7 592 78
GenBank mRNA AJ567756.1 - 3 - 4 202 78
X65551.1 - 6 - 7 593 78

View Full Product Details