Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 148986282-148986806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3TC2
Entrez Gene ID
79628
Gene Name
SH3 domain and tetratricopeptide repeats 2
Gene Aliases
CMT4C, MNMN
UniGene
Hs.483784
Gene Chromosome Location
Chr.5: 148982150-149063174 on Build GRCh38
Transcripts
NM_024577.3

SNP Detail

SNP ID Chromosome Location
rs891414730 Chr.5: 148986349-148986349 on Build GRCh38
rs562519534 Chr.5: 148986537-148986537 on Build GRCh38
rs570361048 Chr.5: 148986741-148986741 on Build GRCh38
rs564549424 Chr.5: 148986556-148986556 on Build GRCh38
rs1042272258 Chr.5: 148986697-148986697 on Build GRCh38
rs916074695 Chr.5: 148986519-148986519 on Build GRCh38
rs529721970 Chr.5: 148986549-148986549 on Build GRCh38
rs141547859 Chr.5: 148986619-148986619 on Build GRCh38
rs748018709 Chr.5: 148986689-148986689 on Build GRCh38
rs558102068 Chr.5: 148986443-148986443 on Build GRCh38
rs1002910738 Chr.5: 148986455-148986455 on Build GRCh38
rs964164055 Chr.5: 148986315-148986315 on Build GRCh38
rs886060102 Chr.5: 148986477-148986477 on Build GRCh38
rs1011761341 Chr.5: 148986696-148986696 on Build GRCh38
rs998263623 Chr.5: 148986392-148986392 on Build GRCh38
rs928979679 Chr.5: 148986631-148986631 on Build GRCh38
rs778554838 Chr.5: 148986654-148986654 on Build GRCh38
rs535418704 Chr.5: 148986398-148986398 on Build GRCh38
rs957894788 Chr.5: 148986760-148986760 on Build GRCh38
rs754723919 Chr.5: 148986525-148986525 on Build GRCh38
rs990684479 Chr.5: 148986526-148986526 on Build GRCh38
rs561010317 Chr.5: 148986766-148986766 on Build GRCh38
rs1036098205 Chr.5: 148986457-148986457 on Build GRCh38
rs1007127777 Chr.5: 148986403-148986403 on Build GRCh38
rs544367145 Chr.5: 148986515-148986515 on Build GRCh38
rs1011730391 Chr.5: 148986417-148986417 on Build GRCh38
rs916474829 Chr.5: 148986542-148986542 on Build GRCh38
rs549872434 Chr.5: 148986764-148986764 on Build GRCh38
rs576698539 Chr.5: 148986458-148986458 on Build GRCh38
rs962520509 Chr.5: 148986563-148986563 on Build GRCh38
rs147903089 Chr.5: 148986412-148986412 on Build GRCh38
rs957631093 Chr.5: 148986460-148986460 on Build GRCh38
rs1020385174 Chr.5: 148986434-148986434 on Build GRCh38
rs147025723 Chr.5: 148986702-148986702 on Build GRCh38
rs865984283 Chr.5: 148986411-148986411 on Build GRCh38
rs572622334 Chr.5: 148986437-148986437 on Build GRCh38
rs912198904 Chr.5: 148986656-148986656 on Build GRCh38
rs148277420 Chr.5: 148986721-148986721 on Build GRCh38
rs973842270 Chr.5: 148986569-148986569 on Build GRCh38
rs1012829733 Chr.5: 148986763-148986763 on Build GRCh38
rs560073983 Chr.5: 148986671-148986671 on Build GRCh38
rs1040000315 Chr.5: 148986684-148986684 on Build GRCh38
rs1050804854 Chr.5: 148986655-148986655 on Build GRCh38
rs1003352111 Chr.5: 148986716-148986716 on Build GRCh38
rs942312637 Chr.5: 148986664-148986664 on Build GRCh38
rs991639575 Chr.5: 148986479-148986479 on Build GRCh38
rs1051193293 Chr.5: 148986346-148986346 on Build GRCh38
rs191187957 Chr.5: 148986754-148986754 on Build GRCh38
rs1048712339 Chr.5: 148986634-148986634 on Build GRCh38
rs901686559 Chr.5: 148986678-148986678 on Build GRCh38
rs546453324 Chr.5: 148986438-148986438 on Build GRCh38

View Full Product Details