Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004066.2 | NP_004057.1 | 1 - 1 | 167 | 70 |
GenBank mRNA | AK312047.1 | - | 1 - 1 | 130 | 70 |
BC029515.1 | - | 1 - 1 | 153 | 70 | |
DB448454.1 | - | 1 - 1 | 167 | 70 | |
U03270.1 | - | 1 - 1 | 143 | 70 |