Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005023.3 | NP_005014.2 | 1 - 2 | 160 | 66 |
XM_005272020.3 | XP_005272077.2 | 1 - 2 | 175 | 66 | |
XM_011543490.2 | XP_011541792.1 | 1 - 2 | 175 | 66 | |
XM_011543492.2 | XP_011541794.1 | 1 - 2 | 168 | 66 | |
GenBank mRNA | AY780790.1 | - | 1 - 2 | 139 | 66 |
DA024020.1 | - | 1 - 2 | 160 | 66 |