Assay Design
Gene Transcripts
- Gene Symbol
- SIRPB1
- Entrez Gene ID
-
10326
- Gene Name
- signal regulatory protein beta 1
- Gene Aliases
-
CD172b, SIRP-BETA-1
- Chromosome Location
-
Chr.20: 1564193 - 1620457 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK313517.1 |
- |
4 - 5 |
1152 |
85 |
Y10376.1 |
- |
4 - 5 |
1127 |
85 |
View Full Product Details