Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 18122056-18122551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NHLRC1
Entrez Gene ID
378884
Gene Name
NHL repeat containing E3 ubiquitin protein ligase 1
Gene Aliases
EPM2A, EPM2B, MALIN, bA204B7.2
UniGene
Hs.348351
Gene Chromosome Location
Chr.6: 18120487-18122620 on Build GRCh38
Transcripts
NM_198586.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4566931 c.512_513CC>TT p.A171V Chr.6: 18122094-18122095 on Build GRCh38
6535869 c.463G>C p.D155H Chr.6: 18122144-18122144 on Build GRCh38
162905 c.540C>T p.V180V Chr.6: 18122067-18122067 on Build GRCh38
6337071 c.463G>T p.D155Y Chr.6: 18122144-18122144 on Build GRCh38
4489761 c.352C>T p.H118Y Chr.6: 18122255-18122255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs912302534 Chr.6: 18122357-18122357 on Build GRCh38
rs1023351397 Chr.6: 18122152-18122152 on Build GRCh38
rs967125703 Chr.6: 18122216-18122216 on Build GRCh38
rs1044313704 Chr.6: 18122406-18122406 on Build GRCh38
rs143893542 Chr.6: 18122496-18122496 on Build GRCh38
rs1011706675 Chr.6: 18122177-18122177 on Build GRCh38
rs75231955 Chr.6: 18122162-18122162 on Build GRCh38
rs568131096 Chr.6: 18122300-18122300 on Build GRCh38
rs750465793 Chr.6: 18122221-18122221 on Build GRCh38
rs370292624 Chr.6: 18122193-18122193 on Build GRCh38
rs776841950 Chr.6: 18122473-18122473 on Build GRCh38
rs749599233 Chr.6: 18122158-18122158 on Build GRCh38
rs986573010 Chr.6: 18122339-18122339 on Build GRCh38
rs764691077 Chr.6: 18122500-18122500 on Build GRCh38
rs903344687 Chr.6: 18122499-18122499 on Build GRCh38
rs886061254 Chr.6: 18122210-18122210 on Build GRCh38
rs781300542 Chr.6: 18122137-18122137 on Build GRCh38
rs763950568 Chr.6: 18122110-18122110 on Build GRCh38
rs757759398 Chr.6: 18122509-18122509 on Build GRCh38
rs886043541 Chr.6: 18122454-18122454 on Build GRCh38
rs755232826 Chr.6: 18122373-18122373 on Build GRCh38
rs764038720 Chr.6: 18122198-18122198 on Build GRCh38
rs756277884 Chr.6: 18122149-18122149 on Build GRCh38
rs762766683 Chr.6: 18122301-18122301 on Build GRCh38
rs200559475 Chr.6: 18122169-18122169 on Build GRCh38
rs754279385 Chr.6: 18122247-18122247 on Build GRCh38
rs779596932 Chr.6: 18122525-18122525 on Build GRCh38
rs758638029 Chr.6: 18122497-18122497 on Build GRCh38
rs769497255 Chr.6: 18122297-18122297 on Build GRCh38
rs759502412 Chr.6: 18122127-18122127 on Build GRCh38
rs201476733 Chr.6: 18122126-18122126 on Build GRCh38
rs754156735 Chr.6: 18122369-18122369 on Build GRCh38
rs187783545 Chr.6: 18122304-18122304 on Build GRCh38
rs779507031 Chr.6: 18122408-18122408 on Build GRCh38
rs774110305 Chr.6: 18122307-18122307 on Build GRCh38
rs963940421 Chr.6: 18122291-18122291 on Build GRCh38
rs769144119 Chr.6: 18122403-18122403 on Build GRCh38
rs909097062 Chr.6: 18122294-18122294 on Build GRCh38
rs767683762 Chr.6: 18122217-18122217 on Build GRCh38
rs750530014 Chr.6: 18122351-18122351 on Build GRCh38
rs765124655 Chr.6: 18122102-18122102 on Build GRCh38
rs759084715 Chr.6: 18122101-18122101 on Build GRCh38
rs200595273 Chr.6: 18122129-18122129 on Build GRCh38
rs748730347 Chr.6: 18122528-18122528 on Build GRCh38
rs761458140 Chr.6: 18122423-18122423 on Build GRCh38
rs796052758 Chr.6: 18122381-18122381 on Build GRCh38
rs766657432 Chr.6: 18122368-18122368 on Build GRCh38
rs778080945 Chr.6: 18122400-18122400 on Build GRCh38
rs774955028 Chr.6: 18122299-18122299 on Build GRCh38
rs772440533 Chr.6: 18122286-18122286 on Build GRCh38
rs115931931 Chr.6: 18122295-18122295 on Build GRCh38
rs933167592 Chr.6: 18122396-18122396 on Build GRCh38
rs148907696 Chr.6: 18122094-18122094 on Build GRCh38
rs758945379 Chr.6: 18122382-18122382 on Build GRCh38
rs950907157 Chr.6: 18122230-18122230 on Build GRCh38
rs758394138 Chr.6: 18122108-18122108 on Build GRCh38
rs529490452 Chr.6: 18122103-18122103 on Build GRCh38
rs143537405 Chr.6: 18122185-18122185 on Build GRCh38
rs756365176 Chr.6: 18122235-18122235 on Build GRCh38
rs760631929 Chr.6: 18122461-18122461 on Build GRCh38
rs148035405 Chr.6: 18122189-18122189 on Build GRCh38
rs370573413 Chr.6: 18122412-18122412 on Build GRCh38
rs757954108 Chr.6: 18122139-18122139 on Build GRCh38
rs587776542 Chr.6: 18122138-18122139 on Build GRCh38
rs10949483 Chr.6: 18122275-18122275 on Build GRCh38
rs780535678 Chr.6: 18122150-18122150 on Build GRCh38
rs759287189 Chr.6: 18122184-18122184 on Build GRCh38
rs866168324 Chr.6: 18122501-18122501 on Build GRCh38
rs770608651 Chr.6: 18122181-18122181 on Build GRCh38
rs753103968 Chr.6: 18122380-18122380 on Build GRCh38
rs765485168 Chr.6: 18122488-18122488 on Build GRCh38
rs759908693 Chr.6: 18122485-18122485 on Build GRCh38
rs28940575 Chr.6: 18122531-18122531 on Build GRCh38
rs1006937839 Chr.6: 18122136-18122136 on Build GRCh38
rs774953311 Chr.6: 18122172-18122172 on Build GRCh38
rs780082503 Chr.6: 18122246-18122246 on Build GRCh38
rs779482781 Chr.6: 18122378-18122378 on Build GRCh38
rs796052754 Chr.6: 18122422-18122422 on Build GRCh38
rs766154920 Chr.6: 18122087-18122087 on Build GRCh38
rs533620472 Chr.6: 18122138-18122138 on Build GRCh38
rs376655413 Chr.6: 18122183-18122183 on Build GRCh38
rs761706418 Chr.6: 18122209-18122209 on Build GRCh38
rs769301934 Chr.6: 18122171-18122171 on Build GRCh38
rs553900705 Chr.6: 18122411-18122411 on Build GRCh38
rs570505924 Chr.6: 18122270-18122270 on Build GRCh38
rs766421414 Chr.6: 18122465-18122465 on Build GRCh38
rs761806425 Chr.6: 18122336-18122336 on Build GRCh38
rs893057832 Chr.6: 18122383-18122383 on Build GRCh38
rs113474848 Chr.6: 18122266-18122266 on Build GRCh38
rs767291745 Chr.6: 18122342-18122342 on Build GRCh38
rs752045674 Chr.6: 18122504-18122504 on Build GRCh38
rs772985160 Chr.6: 18122458-18122458 on Build GRCh38
rs28940576 Chr.6: 18122402-18122402 on Build GRCh38
rs779392254 Chr.6: 18122271-18122271 on Build GRCh38
rs796946441 Chr.6: 18122252-18122252 on Build GRCh38
rs760624567 Chr.6: 18122367-18122367 on Build GRCh38
rs539307365 Chr.6: 18122429-18122429 on Build GRCh38
rs751457173 Chr.6: 18122206-18122206 on Build GRCh38
rs371218834 Chr.6: 18122353-18122353 on Build GRCh38
rs796052755 Chr.6: 18122288-18122288 on Build GRCh38
rs893389047 Chr.6: 18122140-18122140 on Build GRCh38

View Full Product Details