Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282535.1 | NP_001269464.1 | 2 - 2 | 870 | 74 |
NM_013364.5 | NP_037496.4 | 2 - 2 | 870 | 74 | |
GenBank mRNA | AF083116.1 | - | 1 - 1 | 561 | 74 |
AK312299.1 | - | 2 - 2 | 797 | 74 | |
AL136878.1 | - | 2 - 2 | 762 | 74 | |
BC105096.1 | - | 1 - 1 | 554 | 74 | |
BC105098.1 | - | 1 - 1 | 554 | 74 |