Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013940.2 | NP_039228.1 | 1 - 1 | 684 | 62 |
XM_011527911.1 | XP_011526213.1 | 2 - 2 | 641 | 62 | |
GenBank mRNA | BC130360.1 | - | 1 - 1 | 684 | 62 |
BC132969.1 | - | 1 - 1 | 684 | 62 |