Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271707.1 | NP_001258636.1 | 3 - 4 | 653 | 66 |
NM_014160.4 | NP_054879.3 | 4 - 5 | 782 | 66 | |
GenBank mRNA | AF302084.1 | - | 4 - 5 | 66 | |
AK091318.1 | - | 4 - 5 | 727 | 66 | |
AK298463.1 | - | 3 - 4 | 580 | 66 | |
AL834512.1 | - | 1 - 2 | 304 | 66 | |
BC001799.2 | - | 2 - 3 | 491 | 66 | |
BC015715.1 | - | 4 - 5 | 66 |