Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282729.1 | NP_001269658.1 | 3 - 4 | 1027 | 66 |
NM_001282730.1 | NP_001269659.1 | 3 - 4 | 1235 | 66 | |
NM_001282731.1 | NP_001269660.1 | 3 - 4 | 1077 | 66 | |
NM_014860.2 | NP_055675.1 | 3 - 4 | 1022 | 66 | |
XM_005264672.4 | XP_005264729.1 | 3 - 4 | 1224 | 66 | |
GenBank mRNA | AB018307.1 | - | 3 - 4 | 764 | 66 |
AF197954.1 | - | 3 - 4 | 783 | 66 | |
AF224759.1 | - | 2 - 3 | 1612 | 66 | |
AK000329.1 | - | 3 - 4 | 792 | 66 | |
AK000616.1 | - | 3 - 4 | 578 | 66 | |
BC074000.1 | - | 3 - 4 | 764 | 66 | |
CR456983.1 | - | 1 - 2 | 421 | 66 |