Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102653.1 | NP_001096123.1 | 21 - 21 | 4977 | 151 |
XM_005263079.4 | XP_005263136.1 | 21 - 21 | 5080 | 151 | |
XM_011532040.2 | XP_011530342.2 | 21 - 21 | 5081 | 151 | |
XM_011532041.2 | XP_011530343.2 | 21 - 21 | 5078 | 151 | |
XM_011532042.2 | XP_011530344.1 | 21 - 21 | 5002 | 151 | |
GenBank mRNA | AB028969.1 | - | 1 - 1 | 3486 | 152 |