Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_120546.1 | 2 - 2 | 1957 | 120 | |
NR_120547.1 | 3 - 3 | 1615 | 120 | ||
GenBank mRNA | BC002734.2 | - | 1 - 1 | 222 | 120 |
BC096708.1 | - | 1 - 1 | 572 | 120 | |
BC098290.1 | - | 1 - 1 | 572 | 120 | |
BC100673.1 | - | 1 - 1 | 572 | 120 | |
DQ289489.1 | - | 2 - 2 | 1956 | 120 | |
DQ289490.1 | - | 2 - 2 | 1203 | 120 | |
M60614.1 | - | 1 - 1 | 1072 | 120 |