Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006108.3 | NP_006099.2 | 1 - 2 | 510 | 82 |
GenBank mRNA | AB051390.1 | - | 1 - 2 | 266 | 82 |
AK074803.1 | - | 1 - 2 | 456 | 82 | |
AK291780.1 | - | 1 - 2 | 458 | 82 | |
BC136513.1 | - | 1 - 2 | 452 | 82 | |
BC136563.1 | - | 1 - 2 | 452 | 82 | |
DA965056.1 | - | 1 - 2 | 510 | 82 |