Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282615.1 | NP_001269544.1 | 2 - 3 | 444 | 73 |
NM_001282616.1 | NP_001269545.1 | 3 - 4 | 539 | 73 | |
XM_017018865.1 | XP_016874354.1 | 3 - 4 | 494 | 73 | |
GenBank mRNA | BC006440.1 | - | 2 - 3 | 361 | 73 |
BP225850.1 | - | 3 - 4 | 518 | 73 |