Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001775.3 | NP_001766.2 | 4 - 5 | 736 | 97 |
NR_132660.1 | 3 - 4 | 600 | 97 | ||
GenBank mRNA | AK297592.1 | - | 3 - 4 | 432 | 97 |
AK303169.1 | - | 3 - 4 | 590 | 97 | |
AK313268.1 | - | 4 - 5 | 673 | 97 | |
BC007964.2 | - | 4 - 5 | 634 | 97 | |
D84276.1 | - | 4 - 5 | 689 | 97 | |
D84277.1 | - | 3 - 4 | 465 | 97 | |
M34461.1 | - | 4 - 5 | 655 | 97 |