Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155236088-155236593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBA
Entrez Gene ID
2629
Gene Name
glucosylceramidase beta
Gene Aliases
GBA1, GCB, GLUC
UniGene
Hs.282997
Gene Chromosome Location
Chr.1: 155234448-155244862 on Build GRCh38
Transcripts
NM_000157.3 NM_001005741.2 NM_001005742.2 NM_001171811.1 NM_001171812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3474894 c.1072C>T p.P358S Chr.1: 155236397-155236397 on Build GRCh38
6121483 c.1205A>G p.Y402C Chr.1: 155236264-155236264 on Build GRCh38
5754217 c.1080A>C p.K360N Chr.1: 155236389-155236389 on Build GRCh38
4762980 c.1129G>C p.A377P Chr.1: 155236340-155236340 on Build GRCh38
7297270 c.1209C>T p.S403S Chr.1: 155236260-155236260 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1064647 Chr.1: 155236431-155236431 on Build GRCh38
rs772645370 Chr.1: 155236558-155236558 on Build GRCh38
rs76228122 Chr.1: 155236264-155236264 on Build GRCh38
rs142504144 Chr.1: 155236145-155236145 on Build GRCh38
rs573022301 Chr.1: 155236175-155236175 on Build GRCh38
rs794727963 Chr.1: 155236439-155236439 on Build GRCh38
rs748563507 Chr.1: 155236474-155236474 on Build GRCh38
rs765182863 Chr.1: 155236310-155236310 on Build GRCh38
rs186573969 Chr.1: 155236499-155236499 on Build GRCh38
rs956776190 Chr.1: 155236500-155236500 on Build GRCh38
rs540967271 Chr.1: 155236411-155236411 on Build GRCh38
rs761628930 Chr.1: 155236393-155236393 on Build GRCh38
rs143222798 Chr.1: 155236377-155236377 on Build GRCh38
rs915524760 Chr.1: 155236190-155236190 on Build GRCh38
rs878853315 Chr.1: 155236292-155236292 on Build GRCh38
rs11558184 Chr.1: 155236294-155236294 on Build GRCh38
rs939103483 Chr.1: 155236230-155236230 on Build GRCh38
rs1064646 Chr.1: 155236443-155236443 on Build GRCh38
rs948281474 Chr.1: 155236192-155236192 on Build GRCh38
rs74979486 Chr.1: 155236276-155236276 on Build GRCh38
rs550660699 Chr.1: 155236563-155236563 on Build GRCh38
rs398123527 Chr.1: 155236298-155236298 on Build GRCh38
rs761681845 Chr.1: 155236302-155236302 on Build GRCh38
rs370347143 Chr.1: 155236225-155236225 on Build GRCh38
rs776118553 Chr.1: 155236514-155236514 on Build GRCh38
rs866313115 Chr.1: 155236280-155236280 on Build GRCh38
rs747197867 Chr.1: 155236155-155236155 on Build GRCh38
rs552886359 Chr.1: 155236215-155236215 on Build GRCh38
rs754956589 Chr.1: 155236198-155236199 on Build GRCh38
rs121908308 Chr.1: 155236295-155236295 on Build GRCh38
rs924015108 Chr.1: 155236154-155236154 on Build GRCh38
rs763847888 Chr.1: 155236195-155236195 on Build GRCh38
rs749736020 Chr.1: 155236250-155236250 on Build GRCh38
rs780045072 Chr.1: 155236200-155236200 on Build GRCh38
rs374463271 Chr.1: 155236457-155236457 on Build GRCh38
rs769571258 Chr.1: 155236520-155236520 on Build GRCh38
rs752959907 Chr.1: 155236338-155236338 on Build GRCh38
rs781356917 Chr.1: 155236314-155236315 on Build GRCh38
rs760307559 Chr.1: 155236285-155236285 on Build GRCh38
rs756684984 Chr.1: 155236479-155236479 on Build GRCh38
rs771575291 Chr.1: 155236258-155236258 on Build GRCh38
rs762944845 Chr.1: 155236419-155236419 on Build GRCh38
rs781563800 Chr.1: 155236227-155236227 on Build GRCh38
rs78188205 Chr.1: 155236399-155236399 on Build GRCh38
rs76539814 Chr.1: 155236384-155236384 on Build GRCh38
rs878869396 Chr.1: 155236232-155236233 on Build GRCh38
rs878853319 Chr.1: 155236272-155236272 on Build GRCh38
rs778140625 Chr.1: 155236353-155236353 on Build GRCh38
rs532252640 Chr.1: 155236556-155236556 on Build GRCh38
rs398123526 Chr.1: 155236409-155236409 on Build GRCh38
rs779561337 Chr.1: 155236507-155236507 on Build GRCh38
rs771522209 Chr.1: 155236365-155236365 on Build GRCh38
rs749633975 Chr.1: 155236356-155236356 on Build GRCh38
rs181720335 Chr.1: 155236436-155236436 on Build GRCh38
rs773947710 Chr.1: 155236260-155236260 on Build GRCh38
rs989591056 Chr.1: 155236183-155236183 on Build GRCh38
rs560605164 Chr.1: 155236493-155236493 on Build GRCh38
rs2230288 Chr.1: 155236376-155236376 on Build GRCh38
rs121908305 Chr.1: 155236379-155236379 on Build GRCh38
rs777901497 Chr.1: 155236129-155236129 on Build GRCh38
rs121908306 Chr.1: 155236328-155236328 on Build GRCh38
rs772994286 Chr.1: 155236521-155236521 on Build GRCh38
rs552492602 Chr.1: 155236541-155236541 on Build GRCh38
rs771744858 Chr.1: 155236266-155236266 on Build GRCh38
rs121908304 Chr.1: 155236416-155236416 on Build GRCh38
rs149487315 Chr.1: 155236269-155236269 on Build GRCh38
rs78198234 Chr.1: 155236420-155236420 on Build GRCh38
rs976829552 Chr.1: 155236149-155236149 on Build GRCh38
rs1064648 Chr.1: 155236366-155236366 on Build GRCh38
rs753583304 Chr.1: 155236296-155236296 on Build GRCh38
rs75391747 Chr.1: 155236305-155236305 on Build GRCh38
rs568829590 Chr.1: 155236156-155236156 on Build GRCh38
rs760310921 Chr.1: 155236383-155236383 on Build GRCh38
rs768315450 Chr.1: 155236512-155236512 on Build GRCh38
rs75528494 Chr.1: 155236254-155236254 on Build GRCh38
rs989622263 Chr.1: 155236504-155236504 on Build GRCh38
rs77321207 Chr.1: 155236441-155236441 on Build GRCh38
rs751598832 Chr.1: 155236313-155236313 on Build GRCh38
rs80317710 Chr.1: 155236374-155236374 on Build GRCh38
rs757670475 Chr.1: 155236477-155236477 on Build GRCh38
rs79311125 Chr.1: 155236413-155236413 on Build GRCh38
rs747848955 Chr.1: 155236516-155236516 on Build GRCh38
rs554541037 Chr.1: 155236167-155236167 on Build GRCh38
rs781306264 Chr.1: 155236331-155236331 on Build GRCh38
rs75548401 Chr.1: 155236246-155236246 on Build GRCh38
rs1057519358 Chr.1: 155236249-155236249 on Build GRCh38
rs757211442 Chr.1: 155236199-155236199 on Build GRCh38
rs121908309 Chr.1: 155236277-155236277 on Build GRCh38
rs78396650 Chr.1: 155236426-155236426 on Build GRCh38
rs754148803 Chr.1: 155236470-155236470 on Build GRCh38
rs536565557 Chr.1: 155236161-155236161 on Build GRCh38
rs756292503 Chr.1: 155236339-155236339 on Build GRCh38
rs781424692 Chr.1: 155236221-155236221 on Build GRCh38
rs77714449 Chr.1: 155236444-155236444 on Build GRCh38
rs756307975 Chr.1: 155236464-155236464 on Build GRCh38
rs80356765 Chr.1: 155236340-155236340 on Build GRCh38
rs371083513 Chr.1: 155236424-155236424 on Build GRCh38
rs9628662 Chr.1: 155236550-155236550 on Build GRCh38
rs764251205 Chr.1: 155236469-155236469 on Build GRCh38
rs1031110227 Chr.1: 155236480-155236480 on Build GRCh38
rs540038248 Chr.1: 155236213-155236213 on Build GRCh38
rs374306700 Chr.1: 155236367-155236367 on Build GRCh38
rs138498426 Chr.1: 155236245-155236245 on Build GRCh38
rs121908307 Chr.1: 155236261-155236261 on Build GRCh38
rs755021234 Chr.1: 155236326-155236326 on Build GRCh38
rs746523678 Chr.1: 155236511-155236511 on Build GRCh38
rs530006262 Chr.1: 155236404-155236404 on Build GRCh38
rs868591897 Chr.1: 155236318-155236318 on Build GRCh38
rs1018247127 Chr.1: 155236131-155236131 on Build GRCh38
rs758981189 Chr.1: 155236502-155236502 on Build GRCh38

View Full Product Details