Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030636.2 | NP_085139.2 | 2 - 3 | 1521 | 78 |
XM_017012658.1 | XP_016868147.1 | 2 - 3 | 1599 | 78 | |
GenBank mRNA | AB051493.1 | - | 2 - 3 | 1262 | 78 |
AK027386.1 | - | 2 - 3 | 1493 | 78 | |
BC065518.1 | - | 3 - 4 | 1421 | 78 |