Assay Design
Gene Transcripts
- Gene Symbol
- TTTY2B
- Entrez Gene ID
-
100101117
- Gene Name
- testis-specific transcript, Y-linked 2B (non-protein coding)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.Y: 6406244 - 6428444 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_003590.1 |
|
4 - 5 |
520 |
73 |
- Gene Symbol
- TTTY2
- Entrez Gene ID
-
60439
- Gene Name
- testis-specific transcript, Y-linked 2 (non-protein coding)
- Gene Aliases
-
NCRNA00109, TTY2, lINC00109
- Chromosome Location
-
Chr.Y: 9736286 - 9758476 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_001536.2 |
|
4 - 5 |
520 |
73 |
View Full Product Details