Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|---|
RefSeq | NM_001035235.3 | NP_001030312.2 | 2 - 3 | 791 | 57 | |
NR_045586.1 | 2 - 3 | 851 | 57 | |||
NR_045587.1 | 1 - 2 | 309 | 57 | |||
GenBank mRNA | AF092038.1 | - | 2 - 3 | 153 | 57 | |
AF293024.1 | - | LIT1246 SRA |
2 - 3 | 375 | 57 | |
AF293025.1 | - | LIT1250 SRA |
2 - 3 | 375 | 57 | |
AF293026.1 | - | LIT1251 SRA |
2 - 3 | 375 | 57 | |
AF318361.1 | - | 2 - 3 | 792 | 57 | ||
AK024640.1 | - | 1 - 2 | 292 | 57 | ||
AK054960.1 | - | 2 - 3 | 1008 | 57 | ||
BC040043.2 | - | 1 - 2 | 123 | 57 | ||
CX165918.1 | - | 2 - 3 | 413 | 57 | ||
DA311407.1 | - | 1 - 2 | 287 | 57 | ||
DQ286291.1 | - | 2 - 3 | 435 | 57 |