Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145195.1 | NP_001138667.1 | 2 - 3 | 536 | 84 |
NM_001282733.1 | NP_001269662.1 | 2 - 3 | 536 | 84 | |
NM_152725.3 | NP_689938.2 | 2 - 3 | 536 | 84 | |
GenBank mRNA | AK055061.1 | - | 2 - 3 | 484 | 84 |
AK303735.1 | - | 2 - 3 | 221 | 84 | |
BC035118.1 | - | 2 - 3 | 468 | 84 | |
BC047635.1 | - | 2 - 3 | 451 | 84 | |
BC065917.1 | - | 1 - 2 | 280 | 84 | |
BC094700.1 | - | 2 - 3 | 412 | 84 | |
BC117323.1 | - | 1 - 2 | 286 | 84 | |
BC143551.1 | - | 1 - 2 | 285 | 84 |