Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152308.2 | NP_689521.1 | 1 - 2 | 326 | 146 |
GenBank mRNA | AK123764.1 | - | 1 - 2 | 315 | 146 |
BC013040.2 | - | 1 - 2 | 294 | 146 | |
BC022427.1 | - | 1 - 2 | 310 | 146 | |
BC031016.1 | - | 1 - 2 | 314 | 146 |