Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001305163.1 | NP_001292092.1 | 13 - 14 | 2454 | 82 |
NM_001305164.1 | NP_001292093.1 | 13 - 14 | 1833 | 82 | |
NM_133368.2 | NP_588609.1 | 13 - 14 | 1855 | 82 | |
XM_005256220.1 | XP_005256277.1 | 13 - 14 | 2162 | 82 | |
XM_011523427.1 | XP_011521729.1 | 14 - 15 | 1941 | 82 | |
XM_011523428.1 | XP_011521730.1 | 14 - 15 | 2255 | 82 | |
XM_011523430.1 | XP_011521732.1 | 14 - 15 | 1882 | 82 | |
XM_017023844.1 | XP_016879333.1 | 14 - 15 | 2628 | 82 | |
GenBank mRNA | AB075852.1 | - | 13 - 14 | 1775 | 82 |
AK027549.1 | - | 13 - 14 | 2405 | 82 | |
AK124601.1 | - | 5 - 6 | 2772 | 82 | |
AK172845.1 | - | 13 - 14 | 1811 | 82 | |
AL834402.1 | - | 1 - 2 | 59 | 82 | |
BC013173.2 | - | 13 - 14 | 1810 | 82 |