Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 86575843-86576338 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNGB3
Entrez Gene ID
54714
Gene Name
cyclic nucleotide gated channel beta 3
Gene Aliases
ACHM1
UniGene
Hs.154433
Gene Chromosome Location
Chr.8: 86573608-86743675 on Build GRCh38
Transcripts
NM_019098.4 XM_011517138.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
160064 c.2110G>C p.E704Q Chr.8: 86576124-86576124 on Build GRCh38
3703443 c.2344C>G p.Q782E Chr.8: 86575890-86575890 on Build GRCh38
319528 c.2368C>G p.P790A Chr.8: 86575866-86575866 on Build GRCh38
6549505 c.2328C>A p.P776P Chr.8: 86575906-86575906 on Build GRCh38
260173 c.2184_2185insA p.E729fs*3 Chr.8: 86576049-86576050 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974279 Chr.8: 86576291-86576291 on Build GRCh38
rs149271453 Chr.8: 86576104-86576104 on Build GRCh38
rs758606162 Chr.8: 86576054-86576054 on Build GRCh38
rs115178876 Chr.8: 86576228-86576228 on Build GRCh38
rs112573107 Chr.8: 86576075-86576075 on Build GRCh38
rs3735971 Chr.8: 86575986-86575986 on Build GRCh38
rs749307247 Chr.8: 86575867-86575867 on Build GRCh38
rs141691152 Chr.8: 86576128-86576128 on Build GRCh38
rs758914061 Chr.8: 86575919-86575921 on Build GRCh38
rs372823505 Chr.8: 86575872-86575872 on Build GRCh38
rs199570140 Chr.8: 86575934-86575935 on Build GRCh38
rs78239264 Chr.8: 86575926-86575926 on Build GRCh38
rs775381466 Chr.8: 86575893-86575893 on Build GRCh38
rs752973745 Chr.8: 86575940-86575940 on Build GRCh38
rs1033335003 Chr.8: 86576049-86576049 on Build GRCh38
rs976179537 Chr.8: 86576192-86576192 on Build GRCh38
rs943446223 Chr.8: 86576185-86576185 on Build GRCh38
rs754303041 Chr.8: 86575954-86575954 on Build GRCh38
rs377050872 Chr.8: 86576034-86576034 on Build GRCh38
rs773182582 Chr.8: 86576121-86576121 on Build GRCh38
rs371276080 Chr.8: 86576168-86576168 on Build GRCh38
rs773745999 Chr.8: 86576141-86576141 on Build GRCh38
rs751773204 Chr.8: 86576241-86576241 on Build GRCh38
rs771653889 Chr.8: 86576143-86576143 on Build GRCh38
rs759176645 Chr.8: 86576142-86576142 on Build GRCh38
rs772725745 Chr.8: 86575912-86575912 on Build GRCh38
rs772444831 Chr.8: 86575892-86575892 on Build GRCh38
rs762489931 Chr.8: 86576003-86576003 on Build GRCh38
rs965927454 Chr.8: 86576273-86576273 on Build GRCh38
rs201770811 Chr.8: 86575999-86575999 on Build GRCh38
rs764662692 Chr.8: 86575909-86575909 on Build GRCh38
rs776684960 Chr.8: 86575900-86575900 on Build GRCh38
rs750535143 Chr.8: 86575884-86575884 on Build GRCh38
rs758659088 Chr.8: 86575967-86575967 on Build GRCh38
rs760554096 Chr.8: 86575895-86575895 on Build GRCh38
rs762547089 Chr.8: 86575918-86575918 on Build GRCh38
rs754176697 Chr.8: 86576027-86576027 on Build GRCh38
rs3735970 Chr.8: 86576020-86576020 on Build GRCh38
rs999117032 Chr.8: 86575881-86575881 on Build GRCh38
rs749597083 Chr.8: 86576106-86576108 on Build GRCh38
rs761227251 Chr.8: 86575908-86575908 on Build GRCh38
rs371558676 Chr.8: 86576292-86576292 on Build GRCh38
rs748785588 Chr.8: 86576127-86576127 on Build GRCh38
rs746549330 Chr.8: 86576035-86576055 on Build GRCh38
rs146180996 Chr.8: 86576167-86576167 on Build GRCh38
rs748065648 Chr.8: 86575990-86575990 on Build GRCh38
rs944713089 Chr.8: 86575886-86575886 on Build GRCh38
rs267602028 Chr.8: 86575869-86575869 on Build GRCh38
rs750503876 Chr.8: 86576035-86576035 on Build GRCh38
rs190864281 Chr.8: 86575989-86575989 on Build GRCh38
rs780131584 Chr.8: 86576055-86576055 on Build GRCh38
rs763561433 Chr.8: 86576169-86576169 on Build GRCh38
rs769748710 Chr.8: 86575997-86575997 on Build GRCh38
rs745975709 Chr.8: 86575965-86575965 on Build GRCh38
rs778384043 Chr.8: 86575866-86575866 on Build GRCh38
rs756553232 Chr.8: 86575946-86575946 on Build GRCh38
rs769689524 Chr.8: 86576110-86576110 on Build GRCh38
rs768453158 Chr.8: 86575898-86575898 on Build GRCh38
rs3735972 Chr.8: 86575970-86575970 on Build GRCh38
rs770450153 Chr.8: 86576071-86576074 on Build GRCh38
rs779087273 Chr.8: 86575964-86575964 on Build GRCh38
rs747190079 Chr.8: 86576062-86576062 on Build GRCh38
rs777022477 Chr.8: 86576165-86576165 on Build GRCh38
rs777210982 Chr.8: 86575929-86575930 on Build GRCh38
rs764496534 Chr.8: 86575949-86575949 on Build GRCh38
rs774113227 Chr.8: 86576011-86576011 on Build GRCh38
rs746764627 Chr.8: 86575976-86575976 on Build GRCh38
rs757799031 Chr.8: 86576033-86576033 on Build GRCh38
rs758454854 Chr.8: 86575931-86575931 on Build GRCh38
rs757716046 Chr.8: 86575956-86575956 on Build GRCh38
rs759113706 Chr.8: 86576019-86576019 on Build GRCh38
rs765995385 Chr.8: 86576010-86576010 on Build GRCh38

View Full Product Details