Assay Design
Gene Transcripts
- Gene Symbol
- SLC5A3
- Entrez Gene ID
-
6526
- Gene Name
- solute carrier family 5 member 3
- Gene Aliases
-
BCW2, SMIT, SMIT1, SMIT2
- Chromosome Location
-
Chr.21: 34073523 - 34106262 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK055913.1 |
- |
1 - 1 |
1155 |
113 |
View Full Product Details