Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022093.1 | NP_071376.1 | 11 - 12 | 2766 | 74 |
XM_017002048.1 | XP_016857537.1 | 11 - 12 | 2771 | 74 | |
XM_017002049.1 | XP_016857538.1 | 10 - 11 | 2508 | 74 | |
GenBank mRNA | AK127044.1 | - | 11 - 12 | 2766 | 74 |
AK127654.1 | - | 1 - 2 | 186 | 74 | |
AL049689.1 | - | 10 - 11 | 2638 | 74 | |
BC136619.1 | - | 10 - 11 | 2683 | 74 | |
BC144305.1 | - | 9 - 10 | 2152 | 74 |