Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163735.1 | NP_001157207.1 | 23 - 24 | 3370 | 61 |
NM_025109.5 | NP_079385.2 | 19 - 20 | 2770 | 61 | |
XM_011525286.1 | XP_011523588.1 | 26 - 27 | 3057 | 61 | |
XM_011525287.1 | XP_011523589.1 | 26 - 27 | 3051 | 61 | |
XM_011525290.2 | XP_011523592.1 | 24 - 25 | 2919 | 61 | |
XM_017025157.1 | XP_016880646.1 | 26 - 27 | 3031 | 61 | |
XM_017025158.1 | XP_016880647.1 | 25 - 26 | 2964 | 61 | |
XM_017025159.1 | XP_016880648.1 | 24 - 25 | 2925 | 61 | |
XM_017025160.1 | XP_016880649.1 | 26 - 27 | 3057 | 61 | |
XM_017025161.1 | XP_016880650.1 | 24 - 25 | 2849 | 61 | |
GenBank mRNA | AB209035.1 | - | 24 - 25 | 4595 | 61 |
AK304073.1 | - | 23 - 24 | 2721 | 61 | |
AL133017.1 | - | 10 - 11 | 3195 | 61 | |
BC008900.2 | - | 19 - 20 | 2161 | 61 | |
BU543266.1 | - | 5 - 6 | 511 | 61 |