Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138797.2 | NP_620152.1 | 3 - 4 | 588 | 74 |
NR_036556.1 | 3 - 4 | 583 | 74 | ||
XM_006724136.1 | XP_006724199.1 | 3 - 4 | 391 | 74 | |
XM_006724137.1 | XP_006724200.1 | 3 - 4 | 452 | 74 | |
XM_011529880.2 | XP_011528182.1 | 3 - 4 | 367 | 74 | |
XM_017028590.1 | XP_016884079.1 | 3 - 4 | 918 | 74 | |
XM_017028591.1 | XP_016884080.1 | 3 - 4 | 366 | 74 | |
GenBank mRNA | BC014641.1 | - | 3 - 4 | 538 | 74 |
BC066909.1 | - | 3 - 4 | 555 | 74 | |
CD300696.1 | - | - | 482 | 74 | |
CR456471.1 | - | 3 - 4 | 548 | 74 |