Assay Design



Gene Transcripts

Gene Symbol
OIP5
Entrez Gene ID
11339
Gene Name
Opa interacting protein 5
Gene Aliases
5730547N13Rik, CT86, LINT-25, MIS18B, MIS18beta, hMIS18beta
Chromosome Location
Chr.15: 41309268 - 41332621 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007280.1 NP_009211.1 2 - 3 454 66
GenBank mRNA AF025441.1 - 2 - 3 400 66
BC015050.1 - 2 - 3 454 66

View Full Product Details