Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153267.4 | NP_694999.3 | 12 - 13 | 2530 | 66 |
NR_125850.1 | 12 - 13 | 2526 | 66 | ||
GenBank mRNA | AK304531.1 | - | 12 - 13 | 2118 | 66 |
BC015417.2 | - | 4 - 5 | 594 | 66 | |
BC016383.1 | - | 12 - 13 | 2248 | 66 | |
BC063634.1 | - | 12 - 13 | 2507 | 66 | |
BX538309.1 | - | 1 - 2 | 3521 | 66 |