Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015155.2 | NP_055970.1 | 11 - 12 | 1441 | 82 |
XM_005252432.3 | XP_005252489.1 | 11 - 12 | 1450 | 82 | |
XM_005252435.3 | XP_005252492.1 | 11 - 12 | 1234 | 82 | |
XM_011519435.2 | XP_011517737.1 | 11 - 12 | 1285 | 82 | |
XM_017015987.1 | XP_016871476.1 | 10 - 11 | 3490 | 82 | |
XM_017015988.1 | XP_016871477.1 | 10 - 11 | 3488 | 82 | |
XM_017015989.1 | XP_016871478.1 | 10 - 11 | 3490 | 82 | |
XM_017015990.1 | XP_016871479.1 | 11 - 12 | 3009 | 82 | |
XM_017015992.1 | XP_016871481.1 | 6 - 7 | 704 | 82 | |
XM_017015993.1 | XP_016871482.1 | 9 - 10 | 1090 | 82 | |
XM_017015994.1 | XP_016871483.1 | 10 - 11 | 1127 | 82 | |
GenBank mRNA | BC131630.1 | - | 10 - 11 | 1158 | 82 |
BC152443.1 | - | 10 - 11 | 1165 | 82 | |
D86971.2 | - | 10 - 11 | 1165 | 82 |