Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 27546724-27547245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf72
Entrez Gene ID
203228
Gene Name
chromosome 9 open reading frame 72
Gene Aliases
ALSFTD, FTDALS, FTDALS1
UniGene
Hs.493639
Gene Chromosome Location
Chr.9: 27546546-27573866 on Build GRCh38
Transcripts
NM_001256054.2 NM_018325.4

SNP Detail

SNP ID Chromosome Location
rs777032641 Chr.9: 27546910-27546910 on Build GRCh38
rs967510835 Chr.9: 27546915-27546915 on Build GRCh38
rs569838922 Chr.9: 27546903-27546903 on Build GRCh38
rs534344622 Chr.9: 27547149-27547149 on Build GRCh38
rs539764106 Chr.9: 27546930-27546930 on Build GRCh38
rs928440405 Chr.9: 27546967-27546967 on Build GRCh38
rs900022894 Chr.9: 27546883-27546883 on Build GRCh38
rs1053890694 Chr.9: 27547179-27547179 on Build GRCh38
rs1008314408 Chr.9: 27546829-27546829 on Build GRCh38
rs1047133318 Chr.9: 27546782-27546782 on Build GRCh38
rs886063833 Chr.9: 27546974-27546974 on Build GRCh38
rs375528640 Chr.9: 27547100-27547100 on Build GRCh38
rs989223404 Chr.9: 27546786-27546786 on Build GRCh38
rs908173026 Chr.9: 27547174-27547174 on Build GRCh38
rs954793245 Chr.9: 27546757-27546757 on Build GRCh38
rs995987748 Chr.9: 27546958-27546958 on Build GRCh38
rs938892105 Chr.9: 27547124-27547124 on Build GRCh38
rs899991877 Chr.9: 27547154-27547154 on Build GRCh38
rs993031281 Chr.9: 27547140-27547140 on Build GRCh38
rs573825931 Chr.9: 27547064-27547064 on Build GRCh38
rs944090847 Chr.9: 27547086-27547086 on Build GRCh38
rs1030958976 Chr.9: 27546999-27546999 on Build GRCh38
rs554902773 Chr.9: 27547046-27547046 on Build GRCh38
rs191690136 Chr.9: 27547022-27547022 on Build GRCh38
rs769768176 Chr.9: 27547032-27547032 on Build GRCh38
rs956359126 Chr.9: 27547010-27547010 on Build GRCh38
rs890320712 Chr.9: 27546979-27546979 on Build GRCh38
rs887153939 Chr.9: 27546824-27546824 on Build GRCh38
rs13691 Chr.9: 27546892-27546892 on Build GRCh38
rs536808404 Chr.9: 27546977-27546977 on Build GRCh38
rs372145469 Chr.9: 27546784-27546784 on Build GRCh38
rs927539422 Chr.9: 27546878-27546878 on Build GRCh38
rs141012074 Chr.9: 27546767-27546767 on Build GRCh38
rs780309291 Chr.9: 27546863-27546863 on Build GRCh38
rs41272887 Chr.9: 27547178-27547178 on Build GRCh38
rs956390083 Chr.9: 27546778-27546778 on Build GRCh38
rs1041138600 Chr.9: 27547094-27547094 on Build GRCh38
rs1019952191 Chr.9: 27546914-27546914 on Build GRCh38
rs927434314 Chr.9: 27547093-27547093 on Build GRCh38
rs1011625087 Chr.9: 27546911-27546911 on Build GRCh38
rs539123521 Chr.9: 27546890-27546890 on Build GRCh38
rs184151041 Chr.9: 27547172-27547172 on Build GRCh38
rs777272230 Chr.9: 27546799-27546799 on Build GRCh38
rs1008809033 Chr.9: 27546997-27546997 on Build GRCh38
rs981230619 Chr.9: 27546939-27546939 on Build GRCh38
rs969557443 Chr.9: 27547072-27547072 on Build GRCh38
rs544706380 Chr.9: 27547190-27547190 on Build GRCh38
rs9103 Chr.9: 27546830-27546830 on Build GRCh38
rs991400162 Chr.9: 27547002-27547002 on Build GRCh38
rs563194682 Chr.9: 27547212-27547212 on Build GRCh38
rs931251783 Chr.9: 27547036-27547036 on Build GRCh38
rs958553991 Chr.9: 27546972-27546972 on Build GRCh38
rs144871912 Chr.9: 27546886-27546886 on Build GRCh38
rs908558366 Chr.9: 27547071-27547071 on Build GRCh38

View Full Product Details