Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35073751-35074251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCG
Entrez Gene ID
2189
Gene Name
Fanconi anemia complementation group G
Gene Aliases
FAG, XRCC9
UniGene
Hs.591084
Gene Chromosome Location
Chr.9: 35073838-35080016 on Build GRCh38
Transcripts
NM_004629.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4401446 c.1807T>A p.S603T Chr.9: 35074170-35074170 on Build GRCh38
3699672 c.1768C>A p.P590T Chr.9: 35074209-35074209 on Build GRCh38
5559782 c.1766T>C p.L589P Chr.9: 35074211-35074211 on Build GRCh38
1462189 c.1821C>A p.A607A Chr.9: 35074156-35074156 on Build GRCh38
6036225 c.1761-8C>A p.? Chr.9: 35074224-35074224 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916607205 Chr.9: 35073855-35073855 on Build GRCh38
rs536976028 Chr.9: 35073963-35073963 on Build GRCh38
rs758575797 Chr.9: 35074076-35074076 on Build GRCh38
rs541868979 Chr.9: 35074209-35074209 on Build GRCh38
rs908992310 Chr.9: 35073834-35073834 on Build GRCh38
rs374723194 Chr.9: 35074175-35074175 on Build GRCh38
rs771669019 Chr.9: 35074112-35074112 on Build GRCh38
rs747199521 Chr.9: 35074074-35074074 on Build GRCh38
rs773844815 Chr.9: 35074104-35074104 on Build GRCh38
rs772995473 Chr.9: 35074219-35074219 on Build GRCh38
rs546450487 Chr.9: 35073927-35073927 on Build GRCh38
rs148311161 Chr.9: 35074059-35074059 on Build GRCh38
rs17878564 Chr.9: 35073857-35073857 on Build GRCh38
rs950681497 Chr.9: 35074009-35074009 on Build GRCh38
rs370950631 Chr.9: 35074172-35074172 on Build GRCh38
rs1011604982 Chr.9: 35073897-35073897 on Build GRCh38
rs748634605 Chr.9: 35074139-35074139 on Build GRCh38
rs768325047 Chr.9: 35074203-35074203 on Build GRCh38
rs374590435 Chr.9: 35074087-35074087 on Build GRCh38
rs889905535 Chr.9: 35073912-35073912 on Build GRCh38
rs757232381 Chr.9: 35074061-35074061 on Build GRCh38
rs990817225 Chr.9: 35073845-35073845 on Build GRCh38
rs577500778 Chr.9: 35073962-35073962 on Build GRCh38
rs763852498 Chr.9: 35074127-35074127 on Build GRCh38
rs756632525 Chr.9: 35074171-35074171 on Build GRCh38
rs17878854 Chr.9: 35074169-35074169 on Build GRCh38
rs760359166 Chr.9: 35074123-35074123 on Build GRCh38
rs928814452 Chr.9: 35074166-35074166 on Build GRCh38
rs746392434 Chr.9: 35074206-35074206 on Build GRCh38
rs764197648 Chr.9: 35073910-35073910 on Build GRCh38
rs1007380049 Chr.9: 35073794-35073794 on Build GRCh38
rs949414610 Chr.9: 35074192-35074192 on Build GRCh38
rs751616369 Chr.9: 35074007-35074007 on Build GRCh38
rs778912220 Chr.9: 35074064-35074064 on Build GRCh38
rs768333399 Chr.9: 35074086-35074086 on Build GRCh38
rs770455405 Chr.9: 35074101-35074101 on Build GRCh38
rs763090963 Chr.9: 35073890-35073890 on Build GRCh38
rs753741558 Chr.9: 35074060-35074060 on Build GRCh38
rs150203467 Chr.9: 35074136-35074136 on Build GRCh38
rs190050556 Chr.9: 35073942-35073942 on Build GRCh38
rs1045684192 Chr.9: 35073881-35073881 on Build GRCh38
rs938945044 Chr.9: 35073877-35073877 on Build GRCh38
rs780129670 Chr.9: 35074080-35074080 on Build GRCh38
rs564742268 Chr.9: 35073811-35073811 on Build GRCh38
rs761695914 Chr.9: 35074140-35074140 on Build GRCh38
rs766524415 Chr.9: 35074159-35074159 on Build GRCh38
rs765150956 Chr.9: 35074218-35074218 on Build GRCh38
rs748619052 Chr.9: 35074100-35074100 on Build GRCh38
rs146349079 Chr.9: 35073832-35073832 on Build GRCh38
rs750436883 Chr.9: 35074157-35074157 on Build GRCh38
rs776337442 Chr.9: 35074204-35074204 on Build GRCh38
rs557547375 Chr.9: 35073943-35073943 on Build GRCh38
rs760047414 Chr.9: 35073812-35073812 on Build GRCh38
rs771451437 Chr.9: 35074183-35074183 on Build GRCh38
rs397507560 Chr.9: 35074174-35074183 on Build GRCh38
rs758407400 Chr.9: 35074158-35074158 on Build GRCh38
rs775113723 Chr.9: 35074195-35074195 on Build GRCh38
rs755275747 Chr.9: 35074164-35074164 on Build GRCh38
rs377305067 Chr.9: 35074088-35074088 on Build GRCh38
rs532302967 Chr.9: 35074124-35074125 on Build GRCh38
rs758980894 Chr.9: 35074111-35074111 on Build GRCh38
rs763000489 Chr.9: 35074220-35074220 on Build GRCh38
rs35057847 Chr.9: 35074025-35074025 on Build GRCh38
rs142441131 Chr.9: 35073793-35073793 on Build GRCh38
rs535188841 Chr.9: 35073858-35073858 on Build GRCh38
rs185360895 Chr.9: 35073895-35073895 on Build GRCh38
rs1043029775 Chr.9: 35073901-35073901 on Build GRCh38
rs751710424 Chr.9: 35074163-35074163 on Build GRCh38
rs781725105 Chr.9: 35074089-35074089 on Build GRCh38
rs56405035 Chr.9: 35074069-35074069 on Build GRCh38
rs1017386436 Chr.9: 35073800-35073800 on Build GRCh38
rs765086974 Chr.9: 35074155-35074155 on Build GRCh38
rs746613103 Chr.9: 35074200-35074200 on Build GRCh38
rs775000011 Chr.9: 35074117-35074117 on Build GRCh38
rs987500738 Chr.9: 35073781-35073781 on Build GRCh38
rs368098479 Chr.9: 35074176-35074176 on Build GRCh38
rs41274875 Chr.9: 35073818-35073818 on Build GRCh38
rs965879469 Chr.9: 35073960-35073960 on Build GRCh38

View Full Product Details