Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35074285-35074783 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCG
Entrez Gene ID
2189
Gene Name
Fanconi anemia complementation group G
Gene Aliases
FAG, XRCC9
UniGene
Hs.591084
Gene Chromosome Location
Chr.9: 35073838-35080016 on Build GRCh38
Transcripts
NM_004629.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6660479 c.1687C>T p.R563W Chr.9: 35074444-35074444 on Build GRCh38
6289022 c.1760+9C>T p.? Chr.9: 35074362-35074362 on Build GRCh38
3848450 c.1696G>A p.E566K Chr.9: 35074435-35074435 on Build GRCh38
5942254 c.1642C>T p.R548* Chr.9: 35074489-35074489 on Build GRCh38
4530737 c.1723G>A p.E575K Chr.9: 35074408-35074408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006103311 Chr.9: 35074310-35074310 on Build GRCh38
rs748899865 Chr.9: 35074527-35074527 on Build GRCh38
rs867078993 Chr.9: 35074517-35074518 on Build GRCh38
rs368746906 Chr.9: 35074522-35074532 on Build GRCh38
rs775105790 Chr.9: 35074509-35074515 on Build GRCh38
rs17883025 Chr.9: 35074723-35074723 on Build GRCh38
rs750982155 Chr.9: 35074469-35074469 on Build GRCh38
rs752936336 Chr.9: 35074435-35074435 on Build GRCh38
rs549578835 Chr.9: 35074373-35074373 on Build GRCh38
rs886240022 Chr.9: 35074691-35074691 on Build GRCh38
rs754618275 Chr.9: 35074591-35074591 on Build GRCh38
rs982094432 Chr.9: 35074572-35074572 on Build GRCh38
rs778436999 Chr.9: 35074610-35074610 on Build GRCh38
rs182660810 Chr.9: 35074616-35074616 on Build GRCh38
rs779834525 Chr.9: 35074489-35074489 on Build GRCh38
rs564707477 Chr.9: 35074337-35074337 on Build GRCh38
rs768235127 Chr.9: 35074332-35074332 on Build GRCh38
rs750212705 Chr.9: 35074429-35074429 on Build GRCh38
rs759675304 Chr.9: 35074427-35074427 on Build GRCh38
rs45453195 Chr.9: 35074695-35074695 on Build GRCh38
rs200572118 Chr.9: 35074521-35074521 on Build GRCh38
rs919739029 Chr.9: 35074651-35074651 on Build GRCh38
rs17879075 Chr.9: 35074704-35074704 on Build GRCh38
rs1041432580 Chr.9: 35074601-35074601 on Build GRCh38
rs773083883 Chr.9: 35074391-35074391 on Build GRCh38
rs777365150 Chr.9: 35074524-35074524 on Build GRCh38
rs902221264 Chr.9: 35074740-35074740 on Build GRCh38
rs757802309 Chr.9: 35074324-35074324 on Build GRCh38
rs970722372 Chr.9: 35074568-35074568 on Build GRCh38
rs1015944796 Chr.9: 35074317-35074317 on Build GRCh38
rs759587128 Chr.9: 35074537-35074537 on Build GRCh38
rs907901400 Chr.9: 35074584-35074584 on Build GRCh38
rs747659856 Chr.9: 35074638-35074638 on Build GRCh38
rs78602605 Chr.9: 35074330-35074330 on Build GRCh38
rs978951546 Chr.9: 35074547-35074547 on Build GRCh38
rs757636054 Chr.9: 35074448-35074448 on Build GRCh38
rs889420744 Chr.9: 35074410-35074410 on Build GRCh38
rs774426027 Chr.9: 35074536-35074536 on Build GRCh38
rs1040683927 Chr.9: 35074725-35074725 on Build GRCh38
rs779387666 Chr.9: 35074326-35074326 on Build GRCh38
rs372023441 Chr.9: 35074539-35074539 on Build GRCh38
rs528832267 Chr.9: 35074667-35074667 on Build GRCh38
rs1032503600 Chr.9: 35074499-35074499 on Build GRCh38
rs754258080 Chr.9: 35074444-35074444 on Build GRCh38
rs786204205 Chr.9: 35074384-35074384 on Build GRCh38
rs761005679 Chr.9: 35074439-35074439 on Build GRCh38
rs762363538 Chr.9: 35074515-35074516 on Build GRCh38
rs901293008 Chr.9: 35074626-35074626 on Build GRCh38
rs931143206 Chr.9: 35074670-35074670 on Build GRCh38
rs535418091 Chr.9: 35074550-35074550 on Build GRCh38
rs770263417 Chr.9: 35074476-35074479 on Build GRCh38
rs777336987 Chr.9: 35074702-35074702 on Build GRCh38
rs1016213276 Chr.9: 35074712-35074712 on Build GRCh38
rs2237856 Chr.9: 35074683-35074683 on Build GRCh38
rs779403243 Chr.9: 35074454-35074454 on Build GRCh38
rs772450349 Chr.9: 35074461-35074461 on Build GRCh38
rs905765015 Chr.9: 35074722-35074722 on Build GRCh38
rs56406106 Chr.9: 35074693-35074694 on Build GRCh38
rs747842715 Chr.9: 35074340-35074340 on Build GRCh38
rs375560213 Chr.9: 35074518-35074518 on Build GRCh38
rs368539027 Chr.9: 35074338-35074338 on Build GRCh38
rs138855718 Chr.9: 35074442-35074442 on Build GRCh38
rs764950409 Chr.9: 35074658-35074658 on Build GRCh38
rs45537335 Chr.9: 35074493-35074493 on Build GRCh38
rs1024155995 Chr.9: 35074544-35074544 on Build GRCh38
rs770689315 Chr.9: 35074532-35074532 on Build GRCh38
rs56217414 Chr.9: 35074519-35074520 on Build GRCh38
rs556815296 Chr.9: 35074650-35074650 on Build GRCh38
rs762761491 Chr.9: 35074411-35074411 on Build GRCh38
rs200466062 Chr.9: 35074413-35074413 on Build GRCh38

View Full Product Details