Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 22076300-22076806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHEX
Entrez Gene ID
5251
Gene Name
phosphate regulating endopeptidase homolog, X-linked
Gene Aliases
HPDR, HPDR1, HYP, HYP1, LXHR, PEX, XLH
UniGene
Hs.495834
Gene Chromosome Location
Chr.X: 22032327-22251310 on Build GRCh38
Transcripts
NM_000444.5 NM_001282754.1 XM_017029579.1

SNP Detail

SNP ID Chromosome Location
rs865923301 Chr.X: 22076757-22076757 on Build GRCh38
rs370698419 Chr.X: 22076418-22076418 on Build GRCh38
rs978939161 Chr.X: 22076614-22076614 on Build GRCh38
rs933332537 Chr.X: 22076412-22076412 on Build GRCh38
rs770212158 Chr.X: 22076784-22076784 on Build GRCh38
rs1050598557 Chr.X: 22076666-22076666 on Build GRCh38
rs769052597 Chr.X: 22076715-22076715 on Build GRCh38
rs749124424 Chr.X: 22076537-22076537 on Build GRCh38
rs922849110 Chr.X: 22076340-22076340 on Build GRCh38
rs375376231 Chr.X: 22076417-22076417 on Build GRCh38
rs370689912 Chr.X: 22076687-22076687 on Build GRCh38
rs906053371 Chr.X: 22076492-22076492 on Build GRCh38
rs922554151 Chr.X: 22076746-22076746 on Build GRCh38
rs760125505 Chr.X: 22076376-22076377 on Build GRCh38
rs934242477 Chr.X: 22076351-22076351 on Build GRCh38
rs140678356 Chr.X: 22076457-22076457 on Build GRCh38
rs936060620 Chr.X: 22076424-22076424 on Build GRCh38
rs546738664 Chr.X: 22076527-22076527 on Build GRCh38
rs777298207 Chr.X: 22076425-22076425 on Build GRCh38
rs1009937891 Chr.X: 22076544-22076544 on Build GRCh38
rs776983292 Chr.X: 22076758-22076758 on Build GRCh38
rs1035924741 Chr.X: 22076519-22076519 on Build GRCh38
rs1020380458 Chr.X: 22076556-22076556 on Build GRCh38
rs772820253 Chr.X: 22076374-22076374 on Build GRCh38
rs929876242 Chr.X: 22076770-22076770 on Build GRCh38
rs879199639 Chr.X: 22076565-22076565 on Build GRCh38
rs772238348 Chr.X: 22076390-22076390 on Build GRCh38
rs771900963 Chr.X: 22076381-22076381 on Build GRCh38
rs901077990 Chr.X: 22076384-22076384 on Build GRCh38
rs748031736 Chr.X: 22076363-22076363 on Build GRCh38
rs200340509 Chr.X: 22076365-22076365 on Build GRCh38
rs779960753 Chr.X: 22076591-22076591 on Build GRCh38
rs1003909887 Chr.X: 22076644-22076644 on Build GRCh38
rs112329519 Chr.X: 22076704-22076704 on Build GRCh38
rs749100990 Chr.X: 22076375-22076375 on Build GRCh38
rs1057517788 Chr.X: 22076475-22076475 on Build GRCh38
rs775756294 Chr.X: 22076405-22076405 on Build GRCh38
rs758768169 Chr.X: 22076509-22076509 on Build GRCh38
rs190777621 Chr.X: 22076586-22076586 on Build GRCh38
rs1057517787 Chr.X: 22076464-22076464 on Build GRCh38
rs762351671 Chr.X: 22076431-22076431 on Build GRCh38
rs1037767064 Chr.X: 22076441-22076441 on Build GRCh38
rs976002584 Chr.X: 22076329-22076329 on Build GRCh38
rs1045310999 Chr.X: 22076523-22076523 on Build GRCh38
rs765989467 Chr.X: 22076512-22076512 on Build GRCh38
rs373119857 Chr.X: 22076516-22076516 on Build GRCh38
rs151306376 Chr.X: 22076435-22076435 on Build GRCh38
rs748586421 Chr.X: 22076763-22076763 on Build GRCh38
rs886560421 Chr.X: 22076426-22076426 on Build GRCh38
rs910948366 Chr.X: 22076651-22076651 on Build GRCh38
rs1040999044 Chr.X: 22076378-22076378 on Build GRCh38

View Full Product Details