Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 22093899-22094396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHEX
Entrez Gene ID
5251
Gene Name
phosphate regulating endopeptidase homolog, X-linked
Gene Aliases
HPDR, HPDR1, HYP, HYP1, LXHR, PEX, XLH
UniGene
Hs.495834
Gene Chromosome Location
Chr.X: 22032327-22251310 on Build GRCh38
Transcripts
NM_000444.5 NM_001282754.1 XM_011545533.1 XM_017029579.1

SNP Detail

SNP ID Chromosome Location
rs771499864 Chr.X: 22094356-22094356 on Build GRCh38
rs766411751 Chr.X: 22094117-22094117 on Build GRCh38
rs745572252 Chr.X: 22094363-22094363 on Build GRCh38
rs774016247 Chr.X: 22094357-22094357 on Build GRCh38
rs886041496 Chr.X: 22093981-22093981 on Build GRCh38
rs775766235 Chr.X: 22094047-22094047 on Build GRCh38
rs904174507 Chr.X: 22093961-22093961 on Build GRCh38
rs748376198 Chr.X: 22094139-22094139 on Build GRCh38
rs751793274 Chr.X: 22094126-22094145 on Build GRCh38
rs113714114 Chr.X: 22094165-22094165 on Build GRCh38
rs768413746 Chr.X: 22094071-22094071 on Build GRCh38
rs113219043 Chr.X: 22094032-22094032 on Build GRCh38
rs756422601 Chr.X: 22093946-22093946 on Build GRCh38
rs369955446 Chr.X: 22094104-22094104 on Build GRCh38
rs1014836537 Chr.X: 22094121-22094122 on Build GRCh38
rs966775765 Chr.X: 22094254-22094254 on Build GRCh38
rs757256702 Chr.X: 22093977-22093977 on Build GRCh38
rs188090469 Chr.X: 22093958-22093958 on Build GRCh38
rs747331129 Chr.X: 22094070-22094070 on Build GRCh38
rs979845433 Chr.X: 22094299-22094299 on Build GRCh38
rs776124957 Chr.X: 22094077-22094077 on Build GRCh38
rs1015129097 Chr.X: 22094159-22094159 on Build GRCh38
rs746328622 Chr.X: 22094017-22094017 on Build GRCh38
rs770371910 Chr.X: 22094275-22094275 on Build GRCh38
rs772441926 Chr.X: 22094019-22094019 on Build GRCh38
rs886041569 Chr.X: 22094029-22094030 on Build GRCh38
rs895636373 Chr.X: 22094185-22094185 on Build GRCh38
rs766217383 Chr.X: 22093937-22093937 on Build GRCh38
rs886041681 Chr.X: 22094097-22094101 on Build GRCh38
rs933593159 Chr.X: 22093931-22093931 on Build GRCh38
rs267606946 Chr.X: 22094009-22094009 on Build GRCh38
rs867137730 Chr.X: 22093993-22093993 on Build GRCh38
rs200585038 Chr.X: 22094102-22094102 on Build GRCh38
rs147430384 Chr.X: 22094090-22094090 on Build GRCh38
rs777840753 Chr.X: 22093956-22093956 on Build GRCh38
rs923462093 Chr.X: 22094365-22094365 on Build GRCh38
rs752863751 Chr.X: 22093938-22093938 on Build GRCh38
rs183566528 Chr.X: 22094253-22094253 on Build GRCh38
rs761416628 Chr.X: 22094079-22094079 on Build GRCh38
rs142833404 Chr.X: 22094021-22094021 on Build GRCh38
rs1027925302 Chr.X: 22094312-22094312 on Build GRCh38
rs137853268 Chr.X: 22094080-22094080 on Build GRCh38
rs779431331 Chr.X: 22093990-22093990 on Build GRCh38
rs371997551 Chr.X: 22094119-22094119 on Build GRCh38
rs773293894 Chr.X: 22094099-22094099 on Build GRCh38
rs886041223 Chr.X: 22093982-22093982 on Build GRCh38
rs764271513 Chr.X: 22094125-22094126 on Build GRCh38
rs757346784 Chr.X: 22094141-22094155 on Build GRCh38
rs6629449 Chr.X: 22094227-22094227 on Build GRCh38
rs373725747 Chr.X: 22094125-22094125 on Build GRCh38
rs898061673 Chr.X: 22094164-22094164 on Build GRCh38
rs267606945 Chr.X: 22094005-22094005 on Build GRCh38
rs1032268783 Chr.X: 22094314-22094314 on Build GRCh38

View Full Product Details