Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 41474155-41474650 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NYX
Entrez Gene ID
60506
Gene Name
nyctalopin
Gene Aliases
CLRP, CSNB1, CSNB1A, CSNB4, NBM1
UniGene
Hs.302019
Gene Chromosome Location
Chr.X: 41447460-41476414 on Build GRCh38
Transcripts
NM_022567.2 XM_017029709.1

SNP Detail

SNP ID Chromosome Location
rs760297861 Chr.X: 41474411-41474411 on Build GRCh38
rs759518213 Chr.X: 41474385-41474385 on Build GRCh38
rs943712452 Chr.X: 41474549-41474549 on Build GRCh38
rs770193408 Chr.X: 41474358-41474358 on Build GRCh38
rs773164669 Chr.X: 41474266-41474266 on Build GRCh38
rs750321603 Chr.X: 41474188-41474188 on Build GRCh38
rs890791307 Chr.X: 41474198-41474198 on Build GRCh38
rs139601761 Chr.X: 41474324-41474324 on Build GRCh38
rs756209419 Chr.X: 41474600-41474600 on Build GRCh38
rs747098081 Chr.X: 41474248-41474248 on Build GRCh38
rs62637038 Chr.X: 41474562-41474562 on Build GRCh38
rs751166456 Chr.X: 41474191-41474191 on Build GRCh38
rs188293278 Chr.X: 41474213-41474213 on Build GRCh38
rs1012584711 Chr.X: 41474294-41474294 on Build GRCh38
rs11798969 Chr.X: 41474269-41474269 on Build GRCh38
rs750428762 Chr.X: 41474414-41474414 on Build GRCh38
rs867257076 Chr.X: 41474615-41474615 on Build GRCh38
rs756403696 Chr.X: 41474482-41474482 on Build GRCh38
rs940112087 Chr.X: 41474216-41474216 on Build GRCh38
rs766735978 Chr.X: 41474426-41474426 on Build GRCh38
rs62637036 Chr.X: 41474493-41474493 on Build GRCh38
rs781111707 Chr.X: 41474456-41474456 on Build GRCh38
rs750835128 Chr.X: 41474326-41474326 on Build GRCh38
rs774830136 Chr.X: 41474519-41474519 on Build GRCh38
rs541336990 Chr.X: 41474217-41474217 on Build GRCh38
rs752815808 Chr.X: 41474594-41474594 on Build GRCh38
rs63749062 Chr.X: 41474180-41474191 on Build GRCh38
rs62637037 Chr.X: 41474502-41474502 on Build GRCh38
rs956143734 Chr.X: 41474401-41474401 on Build GRCh38
rs776200278 Chr.X: 41474539-41474539 on Build GRCh38
rs768139938 Chr.X: 41474592-41474592 on Build GRCh38
rs749538074 Chr.X: 41474495-41474495 on Build GRCh38
rs766462613 Chr.X: 41474287-41474287 on Build GRCh38
rs369077278 Chr.X: 41474264-41474264 on Build GRCh38
rs894876743 Chr.X: 41474538-41474538 on Build GRCh38
rs375834225 Chr.X: 41474255-41474255 on Build GRCh38
rs372496317 Chr.X: 41474330-41474330 on Build GRCh38
rs751312165 Chr.X: 41474564-41474564 on Build GRCh38
rs750782142 Chr.X: 41474417-41474417 on Build GRCh38
rs748271017 Chr.X: 41474354-41474354 on Build GRCh38
rs62637033 Chr.X: 41474307-41474307 on Build GRCh38
rs560823176 Chr.X: 41474368-41474368 on Build GRCh38
rs749478506 Chr.X: 41474607-41474607 on Build GRCh38
rs758241490 Chr.X: 41474499-41474499 on Build GRCh38
rs527481596 Chr.X: 41474204-41474204 on Build GRCh38
rs143486101 Chr.X: 41474488-41474488 on Build GRCh38
rs745741032 Chr.X: 41474504-41474504 on Build GRCh38
rs758820160 Chr.X: 41474422-41474422 on Build GRCh38
rs758871332 Chr.X: 41474329-41474329 on Build GRCh38
rs866443516 Chr.X: 41474175-41474175 on Build GRCh38
rs763017414 Chr.X: 41474190-41474190 on Build GRCh38
rs766895013 Chr.X: 41474396-41474396 on Build GRCh38
rs774095317 Chr.X: 41474382-41474382 on Build GRCh38
rs1012364938 Chr.X: 41474548-41474548 on Build GRCh38
rs780083539 Chr.X: 41474228-41474228 on Build GRCh38
rs180724602 Chr.X: 41474226-41474226 on Build GRCh38
rs765093922 Chr.X: 41474183-41474183 on Build GRCh38
rs866740256 Chr.X: 41474621-41474621 on Build GRCh38
rs773113302 Chr.X: 41474362-41474362 on Build GRCh38
rs762100670 Chr.X: 41474257-41474257 on Build GRCh38
rs974540920 Chr.X: 41474221-41474221 on Build GRCh38
rs894974127 Chr.X: 41474627-41474627 on Build GRCh38
rs376514245 Chr.X: 41474363-41474363 on Build GRCh38
rs766199959 Chr.X: 41474527-41474527 on Build GRCh38
rs201072571 Chr.X: 41474200-41474200 on Build GRCh38
rs746330892 Chr.X: 41474223-41474223 on Build GRCh38
rs965071108 Chr.X: 41474371-41474371 on Build GRCh38
rs755820494 Chr.X: 41474308-41474308 on Build GRCh38
rs62637035 Chr.X: 41474388-41474388 on Build GRCh38
rs763723615 Chr.X: 41474531-41474531 on Build GRCh38
rs932674244 Chr.X: 41474259-41474259 on Build GRCh38
rs761485619 Chr.X: 41474544-41474544 on Build GRCh38
rs62637032 Chr.X: 41474245-41474245 on Build GRCh38
rs754213194 Chr.X: 41474206-41474206 on Build GRCh38
rs1024588513 Chr.X: 41474589-41474589 on Build GRCh38
rs894950378 Chr.X: 41474277-41474277 on Build GRCh38
rs62637034 Chr.X: 41474346-41474346 on Build GRCh38
rs770982596 Chr.X: 41474375-41474375 on Build GRCh38
rs751972090 Chr.X: 41474429-41474429 on Build GRCh38
rs755532922 Chr.X: 41474437-41474437 on Build GRCh38
rs371445863 Chr.X: 41474623-41474623 on Build GRCh38
rs3810733 Chr.X: 41474296-41474296 on Build GRCh38
rs770778282 Chr.X: 41474496-41474496 on Build GRCh38
rs778001025 Chr.X: 41474605-41474605 on Build GRCh38
rs762965024 Chr.X: 41474268-41474268 on Build GRCh38
rs918230180 Chr.X: 41474555-41474555 on Build GRCh38
rs772093712 Chr.X: 41474516-41474516 on Build GRCh38
rs752006200 Chr.X: 41474554-41474554 on Build GRCh38
rs369485262 Chr.X: 41474214-41474214 on Build GRCh38
rs768551423 Chr.X: 41474353-41474353 on Build GRCh38
rs747565073 Chr.X: 41474332-41474332 on Build GRCh38
rs765226112 Chr.X: 41474390-41474390 on Build GRCh38
rs751115117 Chr.X: 41474293-41474293 on Build GRCh38
rs748302686 Chr.X: 41474474-41474474 on Build GRCh38
rs775085199 Chr.X: 41474405-41474405 on Build GRCh38

View Full Product Details