Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 132078709-132079219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD7
Entrez Gene ID
90167
Gene Name
FERM domain containing 7
Gene Aliases
NYS, NYS1, XIPAN
UniGene
Hs.170776
Gene Chromosome Location
Chr.X: 132076986-132128022 on Build GRCh38
Transcripts
NM_001306193.1 NM_194277.2 XM_017029947.1 XM_017029948.1 XM_017029949.1

SNP Detail

SNP ID Chromosome Location
rs771555307 Chr.X: 132078796-132078796 on Build GRCh38
rs770281308 Chr.X: 132078945-132078945 on Build GRCh38
rs925554511 Chr.X: 132079169-132079169 on Build GRCh38
rs1057483070 Chr.X: 132079179-132079179 on Build GRCh38
rs979704265 Chr.X: 132079150-132079150 on Build GRCh38
rs901917911 Chr.X: 132078906-132078906 on Build GRCh38
rs760039580 Chr.X: 132078944-132078944 on Build GRCh38
rs776876410 Chr.X: 132078943-132078943 on Build GRCh38
rs1004054864 Chr.X: 132078964-132078964 on Build GRCh38
rs750373967 Chr.X: 132078887-132078887 on Build GRCh38
rs149411634 Chr.X: 132078801-132078801 on Build GRCh38
rs569063840 Chr.X: 132079127-132079127 on Build GRCh38
rs763195387 Chr.X: 132078870-132078870 on Build GRCh38
rs777016184 Chr.X: 132078835-132078835 on Build GRCh38
rs759686216 Chr.X: 132078841-132078841 on Build GRCh38
rs775898221 Chr.X: 132078953-132078953 on Build GRCh38
rs376220211 Chr.X: 132078907-132078907 on Build GRCh38
rs756025701 Chr.X: 132078890-132078890 on Build GRCh38
rs1013206878 Chr.X: 132078742-132078742 on Build GRCh38
rs887180776 Chr.X: 132079160-132079160 on Build GRCh38
rs1031406035 Chr.X: 132079007-132079007 on Build GRCh38
rs1021674049 Chr.X: 132078790-132078790 on Build GRCh38
rs1002043036 Chr.X: 132079015-132079015 on Build GRCh38
rs182649942 Chr.X: 132079086-132079086 on Build GRCh38
rs763243213 Chr.X: 132078961-132078961 on Build GRCh38
rs190407853 Chr.X: 132079056-132079056 on Build GRCh38
rs7051368 Chr.X: 132078916-132078916 on Build GRCh38
rs755141737 Chr.X: 132078928-132078928 on Build GRCh38
rs61751728 Chr.X: 132078817-132078817 on Build GRCh38
rs747502543 Chr.X: 132078936-132078936 on Build GRCh38
rs770491140 Chr.X: 132079032-132079032 on Build GRCh38
rs771403134 Chr.X: 132078939-132078939 on Build GRCh38
rs571223039 Chr.X: 132079021-132079021 on Build GRCh38
rs377700609 Chr.X: 132079001-132079001 on Build GRCh38
rs913547607 Chr.X: 132079117-132079117 on Build GRCh38
rs771620923 Chr.X: 132078934-132078934 on Build GRCh38
rs925472063 Chr.X: 132078750-132078750 on Build GRCh38
rs878922754 Chr.X: 132079029-132079029 on Build GRCh38
rs764288995 Chr.X: 132078987-132078987 on Build GRCh38
rs185946711 Chr.X: 132078854-132078854 on Build GRCh38
rs910422741 Chr.X: 132079070-132079070 on Build GRCh38
rs773855252 Chr.X: 132078775-132078775 on Build GRCh38
rs771224602 Chr.X: 132078834-132078834 on Build GRCh38
rs766281336 Chr.X: 132079017-132079017 on Build GRCh38
rs756312864 Chr.X: 132078759-132078759 on Build GRCh38
rs773333393 Chr.X: 132078959-132078960 on Build GRCh38
rs774397248 Chr.X: 132079116-132079116 on Build GRCh38
rs772403008 Chr.X: 132079020-132079020 on Build GRCh38
rs753601085 Chr.X: 132078895-132078895 on Build GRCh38
rs968426085 Chr.X: 132078872-132078872 on Build GRCh38
rs764235664 Chr.X: 132078871-132078871 on Build GRCh38
rs145634694 Chr.X: 132079178-132079178 on Build GRCh38
rs752957017 Chr.X: 132078863-132078863 on Build GRCh38
rs943531530 Chr.X: 132078818-132078818 on Build GRCh38
rs768253456 Chr.X: 132078774-132078774 on Build GRCh38
rs772282277 Chr.X: 132078852-132078852 on Build GRCh38
rs1039567662 Chr.X: 132078897-132078897 on Build GRCh38
rs960454788 Chr.X: 132079061-132079061 on Build GRCh38
rs779741935 Chr.X: 132078891-132078891 on Build GRCh38
rs776146495 Chr.X: 132079045-132079045 on Build GRCh38
rs201656639 Chr.X: 132078997-132078997 on Build GRCh38
rs773585549 Chr.X: 132079020-132079020 on Build GRCh38
rs1047076595 Chr.X: 132079145-132079145 on Build GRCh38
rs777290427 Chr.X: 132078935-132078935 on Build GRCh38
rs943244039 Chr.X: 132079081-132079081 on Build GRCh38

View Full Product Details