Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202941946-202942449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADIPOR1
Entrez Gene ID
51094
Gene Name
adiponectin receptor 1
Gene Aliases
ACDCR1, CGI-45, CGI45, PAQR1, TESBP1A
UniGene
Hs.5298
Gene Chromosome Location
Chr.1: 202940825-202958572 on Build GRCh38
Transcripts
NM_001290553.1 NM_001290557.1 NM_001290629.1 NM_015999.5 XM_017001407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6595236 c.888A>G p.T296T Chr.1: 202942136-202942136 on Build GRCh38
4660041 c.970C>T p.R324C Chr.1: 202942054-202942054 on Build GRCh38
308959 c.834C>T p.G278G Chr.1: 202942190-202942190 on Build GRCh38
6595235 c.921T>C p.A307A Chr.1: 202942103-202942103 on Build GRCh38
1158690 c.846C>T p.T282T Chr.1: 202942178-202942178 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374623170 Chr.1: 202942137-202942137 on Build GRCh38
rs945806165 Chr.1: 202942345-202942345 on Build GRCh38
rs749903174 Chr.1: 202942150-202942150 on Build GRCh38
rs908052599 Chr.1: 202942237-202942237 on Build GRCh38
rs139792596 Chr.1: 202942139-202942139 on Build GRCh38
rs765794028 Chr.1: 202942254-202942254 on Build GRCh38
rs780196089 Chr.1: 202942069-202942069 on Build GRCh38
rs753467350 Chr.1: 202942256-202942256 on Build GRCh38
rs115460336 Chr.1: 202942299-202942299 on Build GRCh38
rs1035816776 Chr.1: 202942250-202942250 on Build GRCh38
rs759593783 Chr.1: 202942200-202942200 on Build GRCh38
rs77591059 Chr.1: 202942275-202942275 on Build GRCh38
rs560007363 Chr.1: 202942178-202942178 on Build GRCh38
rs773052813 Chr.1: 202942198-202942198 on Build GRCh38
rs372159963 Chr.1: 202942189-202942189 on Build GRCh38
rs75801970 Chr.1: 202942272-202942272 on Build GRCh38
rs376776537 Chr.1: 202942234-202942234 on Build GRCh38
rs370017434 Chr.1: 202942264-202942264 on Build GRCh38
rs79011601 Chr.1: 202942274-202942274 on Build GRCh38
rs761830447 Chr.1: 202941995-202941995 on Build GRCh38
rs775693036 Chr.1: 202942216-202942216 on Build GRCh38
rs763154047 Chr.1: 202942217-202942217 on Build GRCh38
rs763241138 Chr.1: 202942093-202942093 on Build GRCh38
rs776748107 Chr.1: 202942018-202942018 on Build GRCh38
rs768482547 Chr.1: 202942187-202942187 on Build GRCh38
rs890090049 Chr.1: 202942312-202942312 on Build GRCh38
rs749520707 Chr.1: 202942251-202942251 on Build GRCh38
rs772204797 Chr.1: 202942008-202942008 on Build GRCh38
rs1010129531 Chr.1: 202942260-202942260 on Build GRCh38
rs758155491 Chr.1: 202942266-202942266 on Build GRCh38
rs868624726 Chr.1: 202942179-202942179 on Build GRCh38
rs564395286 Chr.1: 202942401-202942401 on Build GRCh38
rs376585520 Chr.1: 202942202-202942202 on Build GRCh38
rs765459069 Chr.1: 202942066-202942066 on Build GRCh38
rs779694619 Chr.1: 202942176-202942176 on Build GRCh38
rs764337482 Chr.1: 202942221-202942221 on Build GRCh38
rs12072660 Chr.1: 202942104-202942104 on Build GRCh38
rs751901349 Chr.1: 202942226-202942226 on Build GRCh38
rs976221074 Chr.1: 202942415-202942415 on Build GRCh38
rs778848411 Chr.1: 202942188-202942188 on Build GRCh38
rs549668599 Chr.1: 202942055-202942055 on Build GRCh38
rs772084276 Chr.1: 202942190-202942190 on Build GRCh38
rs774373193 Chr.1: 202941988-202941988 on Build GRCh38
rs542114663 Chr.1: 202942262-202942262 on Build GRCh38
rs781601173 Chr.1: 202942146-202942146 on Build GRCh38
rs2275736 Chr.1: 202942357-202942357 on Build GRCh38
rs1034219417 Chr.1: 202942087-202942087 on Build GRCh38
rs111719484 Chr.1: 202942252-202942252 on Build GRCh38
rs749665579 Chr.1: 202942378-202942379 on Build GRCh38
rs755615066 Chr.1: 202942162-202942162 on Build GRCh38
rs375872003 Chr.1: 202942263-202942263 on Build GRCh38
rs757727437 Chr.1: 202942140-202942140 on Build GRCh38
rs910573237 Chr.1: 202942301-202942301 on Build GRCh38
rs768031659 Chr.1: 202942236-202942236 on Build GRCh38
rs759555652 Chr.1: 202942053-202942053 on Build GRCh38

View Full Product Details