Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 226883516-226884025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSEN2
Entrez Gene ID
5664
Gene Name
presenilin 2
Gene Aliases
AD3L, AD4, CMD1V, PS2, STM2
UniGene
Hs.25363
Gene Chromosome Location
Chr.1: 226870572-226903829 on Build GRCh38
Transcripts
NM_000447.2 NM_012486.2 XM_005273199.3 XM_017001835.1 XM_017001836.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6425822 c.161A>C p.E54A Chr.1: 226883724-226883724 on Build GRCh38
3710617 c.184C>A p.R62S Chr.1: 226883747-226883747 on Build GRCh38
5453446 c.291C>T p.V97V Chr.1: 226883755-226883755 on Build GRCh38
6425823 c.260A>C p.E87A Chr.1: 226883724-226883724 on Build GRCh38
1339320 c.216G>A p.P72P Chr.1: 226883779-226883779 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746664513 Chr.1: 226883768-226883768 on Build GRCh38
rs117724727 Chr.1: 226883584-226883584 on Build GRCh38
rs369622085 Chr.1: 226883778-226883778 on Build GRCh38
rs769025026 Chr.1: 226883926-226883933 on Build GRCh38
rs765144719 Chr.1: 226883766-226883766 on Build GRCh38
rs1295644 Chr.1: 226883676-226883676 on Build GRCh38
rs754836755 Chr.1: 226883760-226883760 on Build GRCh38
rs139332886 Chr.1: 226883728-226883728 on Build GRCh38
rs770934706 Chr.1: 226883948-226883948 on Build GRCh38
rs199808788 Chr.1: 226883891-226883891 on Build GRCh38
rs766446160 Chr.1: 226883757-226883757 on Build GRCh38
rs767091693 Chr.1: 226883969-226883969 on Build GRCh38
rs540463755 Chr.1: 226883594-226883594 on Build GRCh38
rs374399405 Chr.1: 226883555-226883555 on Build GRCh38
rs12057618 Chr.1: 226884002-226884002 on Build GRCh38
rs565637526 Chr.1: 226883932-226883932 on Build GRCh38
rs770414354 Chr.1: 226883941-226883944 on Build GRCh38
rs758086169 Chr.1: 226883825-226883825 on Build GRCh38
rs1045868726 Chr.1: 226883632-226883632 on Build GRCh38
rs534434147 Chr.1: 226883961-226883961 on Build GRCh38
rs763029340 Chr.1: 226883947-226883975 on Build GRCh38
rs957302622 Chr.1: 226883973-226883973 on Build GRCh38
rs761909852 Chr.1: 226883672-226883672 on Build GRCh38
rs201119121 Chr.1: 226883911-226883911 on Build GRCh38
rs921318703 Chr.1: 226883700-226883700 on Build GRCh38
rs772372158 Chr.1: 226883782-226883782 on Build GRCh38
rs751261061 Chr.1: 226883851-226883851 on Build GRCh38
rs199699508 Chr.1: 226883689-226883689 on Build GRCh38
rs370677428 Chr.1: 226883978-226883978 on Build GRCh38
rs969637665 Chr.1: 226883604-226883604 on Build GRCh38
rs754756496 Chr.1: 226883680-226883680 on Build GRCh38
rs778247625 Chr.1: 226883886-226883886 on Build GRCh38
rs751858667 Chr.1: 226883671-226883671 on Build GRCh38
rs747296921 Chr.1: 226883892-226883892 on Build GRCh38
rs779700692 Chr.1: 226883720-226883731 on Build GRCh38
rs1046240 Chr.1: 226883824-226883824 on Build GRCh38
rs1051303953 Chr.1: 226883709-226883709 on Build GRCh38
rs377096583 Chr.1: 226883935-226883935 on Build GRCh38
rs879143602 Chr.1: 226883975-226883975 on Build GRCh38
rs199968912 Chr.1: 226883769-226883769 on Build GRCh38
rs1295643 Chr.1: 226883663-226883663 on Build GRCh38
rs148243400 Chr.1: 226883570-226883570 on Build GRCh38
rs749675208 Chr.1: 226883719-226883719 on Build GRCh38
rs63750048 Chr.1: 226883817-226883817 on Build GRCh38
rs553495122 Chr.1: 226883878-226883878 on Build GRCh38
rs1033298092 Chr.1: 226883950-226883950 on Build GRCh38
rs780738564 Chr.1: 226883856-226883856 on Build GRCh38
rs781669984 Chr.1: 226883927-226883929 on Build GRCh38
rs201407396 Chr.1: 226883662-226883662 on Build GRCh38
rs58973334 Chr.1: 226883748-226883748 on Build GRCh38
rs188598190 Chr.1: 226883729-226883729 on Build GRCh38
rs998683467 Chr.1: 226883949-226883949 on Build GRCh38
rs369268719 Chr.1: 226883687-226883687 on Build GRCh38
rs912747670 Chr.1: 226883888-226883888 on Build GRCh38
rs549378958 Chr.1: 226883812-226883812 on Build GRCh38
rs370626639 Chr.1: 226883975-226883976 on Build GRCh38
rs753721866 Chr.1: 226883679-226883679 on Build GRCh38
rs139972151 Chr.1: 226883771-226883771 on Build GRCh38
rs199652777 Chr.1: 226883678-226883678 on Build GRCh38
rs879236659 Chr.1: 226883636-226883636 on Build GRCh38
rs751342739 Chr.1: 226883940-226883940 on Build GRCh38
rs748397935 Chr.1: 226883781-226883781 on Build GRCh38
rs776842226 Chr.1: 226883814-226883814 on Build GRCh38
rs774588853 Chr.1: 226883953-226883975 on Build GRCh38
rs756899463 Chr.1: 226883855-226883855 on Build GRCh38
rs755514702 Chr.1: 226883859-226883859 on Build GRCh38
rs898714857 Chr.1: 226883990-226883990 on Build GRCh38
rs767577993 Chr.1: 226883740-226883740 on Build GRCh38
rs142546082 Chr.1: 226883770-226883770 on Build GRCh38
rs746463508 Chr.1: 226883698-226883698 on Build GRCh38
rs759641621 Chr.1: 226883969-226883970 on Build GRCh38
rs777396999 Chr.1: 226883688-226883688 on Build GRCh38
rs140501902 Chr.1: 226883774-226883774 on Build GRCh38
rs545470666 Chr.1: 226883683-226883683 on Build GRCh38
rs146894466 Chr.1: 226883725-226883725 on Build GRCh38
rs979496823 Chr.1: 226883831-226883831 on Build GRCh38
rs765023751 Chr.1: 226883818-226883818 on Build GRCh38
rs761470677 Chr.1: 226883968-226883968 on Build GRCh38
rs146718524 Chr.1: 226883857-226883857 on Build GRCh38
rs373392465 Chr.1: 226883864-226883864 on Build GRCh38
rs760961297 Chr.1: 226883799-226883799 on Build GRCh38
rs182893958 Chr.1: 226883640-226883640 on Build GRCh38
rs902963110 Chr.1: 226883927-226883927 on Build GRCh38
rs769031756 Chr.1: 226883775-226883775 on Build GRCh38
rs200801915 Chr.1: 226883863-226883863 on Build GRCh38
rs574813788 Chr.1: 226883585-226883585 on Build GRCh38
rs775935657 Chr.1: 226883717-226883717 on Build GRCh38
rs201261403 Chr.1: 226883746-226883746 on Build GRCh38
rs755175525 Chr.1: 226883658-226883658 on Build GRCh38
rs748572509 Chr.1: 226883869-226883869 on Build GRCh38
rs753871802 Chr.1: 226883759-226883759 on Build GRCh38
rs748576189 Chr.1: 226883934-226883934 on Build GRCh38
rs201272715 Chr.1: 226883755-226883755 on Build GRCh38
rs921254742 Chr.1: 226883836-226883836 on Build GRCh38
rs769784241 Chr.1: 226883701-226883701 on Build GRCh38
rs773925469 Chr.1: 226883943-226883975 on Build GRCh38
rs200610057 Chr.1: 226883899-226883899 on Build GRCh38
rs59683545 Chr.1: 226883682-226883682 on Build GRCh38
rs200350640 Chr.1: 226883842-226883842 on Build GRCh38
rs773522773 Chr.1: 226883785-226883785 on Build GRCh38
rs143501870 Chr.1: 226883712-226883712 on Build GRCh38
rs764253265 Chr.1: 226883666-226883666 on Build GRCh38
rs756137748 Chr.1: 226883677-226883677 on Build GRCh38
rs150400387 Chr.1: 226883747-226883747 on Build GRCh38
rs202133351 Chr.1: 226883768-226883768 on Build GRCh38

View Full Product Details