Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 226895344-226895840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSEN2
Entrez Gene ID
5664
Gene Name
presenilin 2
Gene Aliases
AD3L, AD4, CMD1V, PS2, STM2
UniGene
Hs.25363
Gene Chromosome Location
Chr.1: 226870572-226903829 on Build GRCh38
Transcripts
NM_000447.2 NM_012486.2 XM_005273199.3 XM_017001835.1 XM_017001836.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4232749 c.1312C>T p.L438L Chr.1: 226895445-226895445 on Build GRCh38
4462708 c.1257C>T p.S419S Chr.1: 226895489-226895489 on Build GRCh38
399870 c.1346G>C p.*449S Chr.1: 226895578-226895578 on Build GRCh38
5810869 c.1320T>A p.A440A Chr.1: 226895453-226895453 on Build GRCh38
5810868 c.1221T>A p.A407A Chr.1: 226895453-226895453 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537889666 Chr.1: 226895650-226895650 on Build GRCh38
rs200296479 Chr.1: 226895789-226895789 on Build GRCh38
rs750161150 Chr.1: 226895545-226895545 on Build GRCh38
rs996537968 Chr.1: 226895448-226895448 on Build GRCh38
rs63750666 Chr.1: 226895521-226895521 on Build GRCh38
rs769644784 Chr.1: 226895531-226895531 on Build GRCh38
rs200358385 Chr.1: 226895418-226895418 on Build GRCh38
rs200407641 Chr.1: 226895769-226895769 on Build GRCh38
rs552007162 Chr.1: 226895400-226895400 on Build GRCh38
rs200804459 Chr.1: 226895544-226895544 on Build GRCh38
rs138494303 Chr.1: 226895432-226895432 on Build GRCh38
rs748435174 Chr.1: 226895501-226895501 on Build GRCh38
rs151048692 Chr.1: 226895562-226895562 on Build GRCh38
rs1031898325 Chr.1: 226895755-226895755 on Build GRCh38
rs951840153 Chr.1: 226895653-226895653 on Build GRCh38
rs747074838 Chr.1: 226895511-226895511 on Build GRCh38
rs199547508 Chr.1: 226895540-226895540 on Build GRCh38
rs63750110 Chr.1: 226895548-226895548 on Build GRCh38
rs199676786 Chr.1: 226895526-226895526 on Build GRCh38
rs202067223 Chr.1: 226895767-226895767 on Build GRCh38
rs199571588 Chr.1: 226895784-226895784 on Build GRCh38
rs775164469 Chr.1: 226895467-226895467 on Build GRCh38
rs886046062 Chr.1: 226895651-226895651 on Build GRCh38
rs542892604 Chr.1: 226895790-226895790 on Build GRCh38
rs201424233 Chr.1: 226895549-226895549 on Build GRCh38
rs755273692 Chr.1: 226895397-226895397 on Build GRCh38
rs200164062 Chr.1: 226895582-226895582 on Build GRCh38
rs756609078 Chr.1: 226895494-226895494 on Build GRCh38
rs978078595 Chr.1: 226895738-226895738 on Build GRCh38
rs202239448 Chr.1: 226895673-226895673 on Build GRCh38
rs201399057 Chr.1: 226895599-226895599 on Build GRCh38
rs746930366 Chr.1: 226895437-226895437 on Build GRCh38
rs759282140 Chr.1: 226895461-226895461 on Build GRCh38
rs142772982 Chr.1: 226895535-226895535 on Build GRCh38
rs779830498 Chr.1: 226895612-226895612 on Build GRCh38
rs1028051676 Chr.1: 226895466-226895466 on Build GRCh38
rs200016565 Chr.1: 226895623-226895623 on Build GRCh38
rs749755334 Chr.1: 226895539-226895539 on Build GRCh38
rs555914170 Chr.1: 226895480-226895480 on Build GRCh38
rs201802891 Chr.1: 226895386-226895386 on Build GRCh38
rs1028019225 Chr.1: 226895706-226895706 on Build GRCh38
rs200169735 Chr.1: 226895498-226895498 on Build GRCh38
rs1056270568 Chr.1: 226895366-226895366 on Build GRCh38
rs571949639 Chr.1: 226895788-226895788 on Build GRCh38
rs371315050 Chr.1: 226895452-226895452 on Build GRCh38
rs144289421 Chr.1: 226895796-226895796 on Build GRCh38
rs980133650 Chr.1: 226895811-226895811 on Build GRCh38
rs780297845 Chr.1: 226895388-226895388 on Build GRCh38
rs926478945 Chr.1: 226895694-226895694 on Build GRCh38
rs764998593 Chr.1: 226895464-226895464 on Build GRCh38
rs751025333 Chr.1: 226895474-226895474 on Build GRCh38
rs201922151 Chr.1: 226895536-226895536 on Build GRCh38
rs770838733 Chr.1: 226895440-226895440 on Build GRCh38
rs768312153 Chr.1: 226895627-226895627 on Build GRCh38
rs571670427 Chr.1: 226895682-226895682 on Build GRCh38
rs769663194 Chr.1: 226895605-226895605 on Build GRCh38
rs766859560 Chr.1: 226895495-226895495 on Build GRCh38
rs200427880 Chr.1: 226895472-226895472 on Build GRCh38
rs199796978 Chr.1: 226895484-226895484 on Build GRCh38
rs376697912 Chr.1: 226895396-226895396 on Build GRCh38
rs776603544 Chr.1: 226895525-226895525 on Build GRCh38
rs371693894 Chr.1: 226895585-226895585 on Build GRCh38
rs777716175 Chr.1: 226895507-226895507 on Build GRCh38
rs772105831 Chr.1: 226895420-226895420 on Build GRCh38
rs774892527 Chr.1: 226895797-226895797 on Build GRCh38
rs917826465 Chr.1: 226895411-226895411 on Build GRCh38
rs1050239559 Chr.1: 226895815-226895815 on Build GRCh38
rs200745703 Chr.1: 226895597-226895597 on Build GRCh38
rs945284547 Chr.1: 226895722-226895722 on Build GRCh38
rs113600895 Chr.1: 226895377-226895377 on Build GRCh38
rs550113178 Chr.1: 226895652-226895652 on Build GRCh38
rs200943229 Chr.1: 226895660-226895660 on Build GRCh38
rs917857514 Chr.1: 226895717-226895717 on Build GRCh38
rs773754382 Chr.1: 226895537-226895537 on Build GRCh38
rs186288674 Chr.1: 226895711-226895711 on Build GRCh38
rs199791100 Chr.1: 226895796-226895796 on Build GRCh38
rs201559102 Chr.1: 226895772-226895772 on Build GRCh38
rs774092766 Chr.1: 226895631-226895631 on Build GRCh38
rs1006862439 Chr.1: 226895677-226895677 on Build GRCh38
rs143059995 Chr.1: 226895699-226895699 on Build GRCh38
rs758586194 Chr.1: 226895505-226895505 on Build GRCh38
rs201494033 Chr.1: 226895475-226895475 on Build GRCh38
rs987069930 Chr.1: 226895471-226895471 on Build GRCh38
rs201982084 Chr.1: 226895468-226895468 on Build GRCh38
rs775252540 Chr.1: 226895533-226895533 on Build GRCh38
rs144318314 Chr.1: 226895516-226895516 on Build GRCh38
rs201488196 Chr.1: 226895795-226895795 on Build GRCh38
rs758582618 Chr.1: 226895570-226895570 on Build GRCh38
rs201163703 Chr.1: 226895721-226895721 on Build GRCh38
rs778005647 Chr.1: 226895574-226895574 on Build GRCh38

View Full Product Details