Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 6011513-6012052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL2RA
Entrez Gene ID
3559
Gene Name
interleukin 2 receptor subunit alpha
Gene Aliases
CD25, IDDM10, IL2R, IMD41, TCGFR, p55
UniGene
Hs.231367
Gene Chromosome Location
Chr.10: 6010694-6062370 on Build GRCh38
Transcripts
NM_000417.2 NM_001308242.1 NM_001308243.1

SNP Detail

SNP ID Chromosome Location
rs1014045828 Chr.10: 6011754-6011754 on Build GRCh38
rs1005611743 Chr.10: 6011567-6011567 on Build GRCh38
rs886047076 Chr.10: 6011988-6011988 on Build GRCh38
rs544896196 Chr.10: 6011886-6011886 on Build GRCh38
rs55885853 Chr.10: 6011811-6011811 on Build GRCh38
rs760142892 Chr.10: 6011987-6011987 on Build GRCh38
rs12722716 Chr.10: 6011984-6011984 on Build GRCh38
rs1015233762 Chr.10: 6011569-6011569 on Build GRCh38
rs923906350 Chr.10: 6011687-6011687 on Build GRCh38
rs745755601 Chr.10: 6011752-6011752 on Build GRCh38
rs149796519 Chr.10: 6011803-6011803 on Build GRCh38
rs572589682 Chr.10: 6011828-6011828 on Build GRCh38
rs987042934 Chr.10: 6011612-6011612 on Build GRCh38
rs147669116 Chr.10: 6011983-6011983 on Build GRCh38
rs1019307368 Chr.10: 6012016-6012016 on Build GRCh38
rs527536271 Chr.10: 6011622-6011622 on Build GRCh38
rs895143394 Chr.10: 6011742-6011742 on Build GRCh38
rs945508248 Chr.10: 6011656-6011656 on Build GRCh38
rs115247277 Chr.10: 6011611-6011611 on Build GRCh38
rs1570538 Chr.10: 6011605-6011605 on Build GRCh38
rs543983290 Chr.10: 6011713-6011714 on Build GRCh38
rs939943430 Chr.10: 6011907-6011907 on Build GRCh38
rs527589024 Chr.10: 6011909-6011909 on Build GRCh38
rs34164172 Chr.10: 6011864-6011864 on Build GRCh38
rs1055214907 Chr.10: 6011857-6011857 on Build GRCh38
rs775924333 Chr.10: 6011943-6011943 on Build GRCh38
rs916647121 Chr.10: 6011995-6011995 on Build GRCh38
rs759899126 Chr.10: 6011906-6011906 on Build GRCh38
rs1003974783 Chr.10: 6011796-6011796 on Build GRCh38
rs370765858 Chr.10: 6011638-6011638 on Build GRCh38
rs141524746 Chr.10: 6011831-6011831 on Build GRCh38
rs151183621 Chr.10: 6011548-6011548 on Build GRCh38
rs542812308 Chr.10: 6011830-6011830 on Build GRCh38
rs529774081 Chr.10: 6011557-6011557 on Build GRCh38
rs12722718 Chr.10: 6011848-6011848 on Build GRCh38
rs571299400 Chr.10: 6011797-6011797 on Build GRCh38
rs925329636 Chr.10: 6011688-6011688 on Build GRCh38
rs992186765 Chr.10: 6011988-6011988 on Build GRCh38
rs936715109 Chr.10: 6011856-6011856 on Build GRCh38
rs936785292 Chr.10: 6011698-6011698 on Build GRCh38
rs962737707 Chr.10: 6011801-6011801 on Build GRCh38
rs28360484 Chr.10: 6011846-6011846 on Build GRCh38
rs533020456 Chr.10: 6011595-6011595 on Build GRCh38
rs985341613 Chr.10: 6011643-6011643 on Build GRCh38
rs537968636 Chr.10: 6011772-6011772 on Build GRCh38
rs953874339 Chr.10: 6011639-6011639 on Build GRCh38
rs967967407 Chr.10: 6011665-6011665 on Build GRCh38
rs997842886 Chr.10: 6011876-6011876 on Build GRCh38
rs781076912 Chr.10: 6011825-6011825 on Build GRCh38
rs12722717 Chr.10: 6011919-6011919 on Build GRCh38
rs567205124 Chr.10: 6011696-6011696 on Build GRCh38
rs12722602 Chr.10: 6011903-6011903 on Build GRCh38
rs1046396172 Chr.10: 6011895-6011895 on Build GRCh38
rs374118462 Chr.10: 6011787-6011787 on Build GRCh38
rs763727948 Chr.10: 6011540-6011540 on Build GRCh38
rs968296462 Chr.10: 6011944-6011944 on Build GRCh38
rs12722603 Chr.10: 6011686-6011686 on Build GRCh38
rs896837610 Chr.10: 6011602-6011602 on Build GRCh38
rs949376143 Chr.10: 6011865-6011865 on Build GRCh38
rs912803984 Chr.10: 6011838-6011838 on Build GRCh38
rs1022617157 Chr.10: 6011658-6011658 on Build GRCh38

View Full Product Details