Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015286.5 | NP_056101.5 | 2 - 3 | 1059 | 92 |
NM_145728.2 | NP_663780.2 | 2 - 3 | 1059 | 92 | |
XM_017022035.1 | XP_016877524.1 | 2 - 3 | 1070 | 92 | |
GenBank mRNA | AB002351.2 | - | 2 - 3 | 1097 | 92 |
AF359284.1 | - | 2 - 3 | 1050 | 92 | |
AJ310521.1 | - | 2 - 3 | 1053 | 92 | |
AJ310522.1 | - | 2 - 3 | 1053 | 92 | |
AJ697971.1 | - | 2 - 3 | 1059 | 92 | |
AL833262.1 | - | 2 - 3 | 550 | 92 | |
BC151243.1 | - | 2 - 3 | 1104 | 92 |