Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014992.2 | NP_055807.1 | 14 - 15 | 2010 | 66 |
XM_005267430.1 | XP_005267487.1 | 15 - 16 | 2168 | 66 | |
XM_005267431.1 | XP_005267488.1 | 15 - 16 | 2268 | 66 | |
GenBank mRNA | AB014566.1 | - | 14 - 15 | 1996 | 66 |
AK021890.1 | - | 4 - 5 | 433 | 66 |