Assay Design



Gene Transcripts

Gene Symbol
SPON1
Entrez Gene ID
10418
Gene Name
spondin 1
Gene Aliases
VSGP/F-spondin, f-spondin
Chromosome Location
Chr.11: 13962637 - 14268133 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006108.3 NP_006099.2 7 - 8 1159 76
GenBank mRNA AB018305.2 - 6 - 7 644 76
AB051390.1 - 7 - 8 915 76
AK291780.1 - 7 - 8 1107 76

View Full Product Details